Skip to Content
Merck
All Photos(2)

Key Documents

318701

Sigma-Aldrich

Bromocresol Green sodium salt solution

0.04 wt. % in H2O

Synonym(s):

3′,3′′,5′,5′′-Tetrabromo-m-cresolsulfonephthalein sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€82.90

€82.90


Estimated to ship on06 April 2025


Request a Bulk Order

Select a Size

Change View
500 ML
€82.90

About This Item

Empirical Formula (Hill Notation):
C21H13Br4NaO5S
CAS Number:
Molecular Weight:
720.00
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€82.90


Estimated to ship on06 April 2025


Request a Bulk Order

form

liquid

Quality Level

concentration

0.04 wt. % in H2O

visual transition interval

3.8-5.4, yellow to blue

density

0.981 g/mL at 25 °C

λmax

612 nm

ε (extinction coefficient)

≥10000 at 306-312 nm
≥8000 at 397-403 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Cc1c(Br)c(O)c(Br)cc1C2(OS(=O)(=O)c3ccccc23)c4cc(Br)c([O-])c(Br)c4C

InChI

1S/C21H14Br4O5S.Na/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21;/h3-8,26-27H,1-2H3;/q;+1/p-1

InChI key

HEFSGAHJDGZCHA-UHFFFAOYSA-M

Biochem/physiol Actions

Bromocresol green is a sulfonephthalein dye.[1] It is mainly used for measuring serum albumin. In acidic conditions, bromocresol green interacts with albumin, thereby resulting in an increase in absorbance at 630nm. The change in absorbance is linked with the concentration of albumin.[2] Bromocresol Green is also a pH indicator, changing from yellow (pH 3.8) to blue color (5.4).[3] It is widely used as a colloidal pigment.[1]

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David B. Troy, Paul Beringer
Remington: The Science and Practice of Pharmacy (2006)
Homocysteine as a predictive biomarker in early diagnosis of renal failure susceptibility and prognostic diagnosis for end stages renal disease.
Amin HK, et.al.
Renal Failure, 38, 1267-1275 (2016)
Homocysteine as a predictive biomarker in early diagnosis of renal failure susceptibility and prognostic diagnosis for end stages renal disease.
Amin HK, et al.
Renal Failure, 38, 1267-1275 (2016)
R. W. Sabnis
Handbook of Acid-Base Indicators (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service