Skip to Content
Merck
All Photos(3)

Key Documents

114375

Sigma-Aldrich

Bromocresol Purple

BioReagent, suitable for indicator, Dye content 90 %

Synonym(s):

5,5′-Dibromo-o-cresolsulfonphthalein, Bromcresol purple sultone form

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€68.90
25 G
€204.00

€68.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€68.90
25 G
€204.00

About This Item

Empirical Formula (Hill Notation):
C21H16Br2O5S
CAS Number:
Molecular Weight:
540.22
Beilstein:
359618
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€68.90


Check Cart for Availability

Request a Bulk Order

product line

BioReagent

Quality Level

Assay

≥89.0% (HPLC)

form

powder

composition

Dye content, 90%

technique(s)

titration: suitable

pH

5.2-6.8, yellow to purple

visual transition interval

5.2-6.8, yellow to blue

mp

240 °C (dec.) (lit.)

solubility

ethanol: 1 mg/mL

λmax

419 nm

ε (extinction coefficient)

≥22000 at 418-424 nm in methanol

suitability

suitable for indicator

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1cc(cc(Br)c1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C)c(O)c(Br)c4

InChI

1S/C21H16Br2O5S/c1-11-7-13(9-16(22)19(11)24)21(14-8-12(2)20(25)17(23)10-14)15-5-3-4-6-18(15)29(26,27)28-21/h3-10,24-25H,1-2H3

InChI key

ABIUHPWEYMSGSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bromocresol Purple has been used for staining of Saccharomyces cerevisiae.[1]
Spectrophotometric determination of serum-albumin

Biochem/physiol Actions

Bromocresol purple is a pH indicator which changes color from yellow (at low pH 5.2) to violet (above pH 6.8).[2] It is mainly used as a fluorescent stain in yeast cells. It helps in dead cell count for cells with plasma membrane damage. Cells with proper cell membrane are not stained, while damaged cells are seen as blue-grey ghosts.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescent staining with bromocresol purple: a rapid method for determining yeast cell dead count developed as an assay of killer toxin activity.
Kurzweilova H and Sigler K
Yeast, 9, 1207-1211 (1993)
ATP leakage from yeast cells treated by extracellular glycolipids of Pseudozyma fusiformata.
Kulakovskaya TV, et al.
FEMS Yeast Research, 3, 401-404 (2003)
Yeast colonies synchronise their growth and development.
Palkova Z and Forstova J
Journal of Cell Science, 113, 1923-1928 (2000)
Munzer Kattan et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 69(2), 377-380 (2010-11-27)
Polyvinyl chloride (PVC) dyed with bromocresol purple was investigated as a high-dose radiation dosimeter. The absorbance at 417 nm depends linearly on the dose below 50 kGy. The response depends neither on dose rate nor on the irradiation temperature. The
Faizah Mohd Faizul et al.
Journal of photochemistry and photobiology. B, Biology, 90(1), 1-7 (2007-11-21)
The interaction between bromocresol purple (BCP) and bovine serum albumin (BSA)/porcine serum albumin (PSA) was investigated both in the absence and presence of bilirubin (BR) using absorption/absorption difference spectroscopy. A significant red shift in the absorption maxima of BCP accompanied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service