Skip to Content
Merck
All Photos(3)

Key Documents

105945

Sigma-Aldrich

Phenolphthalein

Phenolphthalein
1 of 1 reviewers received a sample product or took part in a promotion

ACS reagent

Synonym(s):

3,3-Bis(4-hydroxyphenyl)-1(3H)-isobenzofuranone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14O4
CAS Number:
Molecular Weight:
318.32
Colour Index Number:
764
Beilstein:
284423
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

form

powder

technique(s)

titration: suitable

visual transition interval

8.0-10, colorless to red

mp

261-263 °C (lit.)

solubility

acetone: soluble

density

1.27 g/cm3 at 32 °C

λmax

374 nm (2nd)
552 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc(cc1)C2(OC(=O)c3ccccc23)c4ccc(O)cc4

InChI

1S/C20H14O4/c21-15-9-5-13(6-10-15)20(14-7-11-16(22)12-8-14)18-4-2-1-3-17(18)19(23)24-20/h1-12,21-22H

InChI key

KJFMBFZCATUALV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenolphthalein is an indicator for acid-base titration reactions.[1] It changes from colorless to violet-red between pH 8.3-9.8, it is red in extremely acidic solutions, and colorless in extremely alkaline solutions.[2]

It is present as a nonionic, lipophilic lactone in aqueous neutral solutions. It exists as a hydrophilic dianion in alkaline solutions.

Application

Phenolphthalein has been commonly used as an indicator in the fields of biology and medicine. Its applications include:

  • the development of novel technique to monitor the vacuum freeze-drying process[3]
  • the determination of the depth of carbonation in concrete[4]
  • the utilization of liquid marbles to sense gaseous environmental components
  • the estimation of total acidity in vinegar
  • the assessment of sodium hydroxide penetration into potatoes following chemical peeling
  • as a model organic compound in the evaluation of the capture efficiency of various cyclodextrin constructs such as nanofiber-CD

Biochem/physiol Actions

Phenolphthalein is colorless in acidic solutions and turns pink in basic solutions.

Principle

Phenolphthalein undergoes structural modification when there is a change in pH or in the structural state of water.[1] It is a nonionic, lipophilic colorless lactone in aqueous neutral solutions and a hydrophilic dianion in alkaline solutions.[2]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Teaching New Tricks to an Old Indicator: pH-Switchable, Photoactive Microporous Polymer Networks from Phenolphthalein with Tunable CO2 Adsorption Power
Kiskan, et al.
Macromolecules (2012)
The experimental investigation of concrete carbonation depth
Cheng-Feng Chang, Jing-Wen Chen
Cement and Concrete Research, 36(9) (2006)
Using phenolphthalein as a promising indicator to monitor the vacuum freeze-drying process
Liu, et al.
Materials Letters, 139 (2015)
Conn?s Biological Stains: A Handbook of Dyes, Stains and Fluorochromes for Use in Biology and Medicine (10th ed.)
RW Horobin, JW Kiernan (Eds.)
Conn?s Biological Stains (2002)
Min Guk Kim et al.
Journal of personalized medicine, 10(4) (2020-10-15)
Unbalanced dietary habits and the consumption of high protein and instant foods cause an increase in constipation. Here, we evaluated the effects of galacto-oligosaccharide (GOS) on a rat model of loperamide-induced constipation by measuring various biological markers and cecal microbiota.

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

    • Review 1
    • Votes 0
    5 out of 5 stars.

    cool indicator!

    really cool indicator! definitely would recommend to friend!

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service