Skip to Content
Merck
All Photos(2)

Key Documents

318752

Sigma-Aldrich

Bromothymol Blue sodium salt solution

0.04 wt. % in H2O

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€28.60

€28.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€28.60

About This Item

Empirical Formula (Hill Notation):
C27H27Br2NaO5S
CAS Number:
Molecular Weight:
646.36
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€28.60


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

concentration

0.04 wt. % in H2O

pH

6.0-7.6, yellow to blue

density

0.99 g/mL at 25 °C (lit.)

λmax

615 nm

ε (extinction coefficient)

≥13000 at 304-310 nm in 0.1 M NaOH at 0.005 g/L
≥8000 at 391-397 nm in 0.1 M NaOH at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CC(C)c1cc(c(C)c(Br)c1O)\C(=C2/C=C(C(C)C)C(=O)C(Br)=C2C)c3ccccc3S([O-])(=O)=O

InChI

1S/C27H28Br2O5S.Na/c1-13(2)18-11-20(15(5)24(28)26(18)30)23(17-9-7-8-10-22(17)35(32,33)34)21-12-19(14(3)4)27(31)25(29)16(21)6;/h7-14,30H,1-6H3,(H,32,33,34);/q;+1/p-1/b23-21+;

InChI key

NMKFVGALBGZKGW-FKWCIMQXSA-M

General description

Bromothymol Blue is a sulfonated hydroxyquinone in acidic solutions. In alkaline solutions, it is present as a dianion that is blue with a larger conjugated system than the yellow hydroxyquinone. Both forms are lipophilic. It is also known as bromthymol blue or dibromothymolsulfonphthalein.

Application

Bromothymol blue has various applications in biology and medicine. It is suitable for demonstrating fungal hyphae within plant roots. It is used as a vital stain to trace the movement of fluids from the lymph. It is added to an agar gel microbial culture medium for enumeration of Bacillus cereus. Bromothymol blue is also used as a pH indicator due to its precise color transition properties at neutrality.

Biochem/physiol Actions

Bromothymol Blue is a pH indicator which changes color from yellow (pH 6.0) to blue (7.6). It belongs to the class of sulfonephthalein dyes.[1]

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R. W. Sabnis
Handbook of Acid-Base Indicators (2007)

Questions

  1. Do you provide dyes or other solutions used for validating adequate cleaning of equipment?

    1 answer
    1. If you are checking for the adequate cleaning of equipment, there are various ways to do so. pH indicators such as bromothymol blue can be used to ensure that alkaline detergents have been washed away. However, dyes are generally not commonly used to check for sterility or whether a surface is clean of all substances. The use of other dyes may vary, and it's essential to consider the specific use and characteristics of each dye for different applications.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service