Skip to Content
Merck

Skip To

MABE243-I

Anti-Histone H3. X/Y Antibody, clone 8H6-2111

clone 8H6-2111, from rat

Synonym(s):

Histone H3 X/Y, H3 X/Y

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
8H6-2111, monoclonal
Species reactivity:
human
Application:
ICC
Citations:
1
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8H6-2111, monoclonal

species reactivity

human

species reactivity (predicted by homology)

primate (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

isotype

IgG2aκ

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... H3Y1(391769)

General description

17 kDa calculated.
Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. It is a core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. Both H3.X and H3.Y are two novel primate-specific Histone H3 variants that are not found other mammals or lower eukaryotes. H3.X and H3.Y mRNAs are expressed in transformed cell lines, such as U2OS, and in low amounts in human bone, breast and lung tissue. They exhibit amino acid sequence homology with other H3 variants, but differ in several functionally important residues, such as serine10 (S to A) and Serine 28 (S to R). Knockdown of H3.Y variant is shown to affect cell growth and leads to changes in the expression of many genes involved in cell cycle control. It is noteworthy that the number of cells expressing H3.Y is increased under stress stimuli.(Ref.: Wiedemann, SM et al. (2010). J. Cell Biol. 90(5); 777-791).

Immunogen

Ovalbumin-conjugated linear peptide corresponding to a sequence of 12 amino acids from the N-terminal region of human Histone H3.X/Y.

Application

Anti-Histone H3. X/Y Antibody, clone 8H6-2111, Cat. No. MABE243-I, is a highly specific rat monoclonal antibody that targets Histone H3. X/Y and has been tested in Immunocytochemistry.
Research Category
Epigenetics & Nuclear Function

Biochem/physiol Actions

Clone 8H6-2111 specifically detects X and Y variants of Histone H3 in primates. It does not recognize HA-H3.1, -H3.2, or -H3.3. This clone targets an epitope of 12 amino acids in the N-terminal region.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Immunocytochemistry in U2OS cells with and without serum starvation.

Immunocytochemistry Analysis: A 1:250 dilution of this antibody detected Histone H3. X/Y in U2OS cells with and without serum starvation.

Other Notes

Concentration: Please refer to lot specific datasheet.
Replaces: MABE243

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
06-75507-523ABE44-S
clone

8H6-2111, monoclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

species reactivity

human

species reactivity

rat, mouse, human

species reactivity

human

species reactivity

human, mouse

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

serum

antibody form

affinity isolated antibody

Gene Information

human ... H3Y1(391769)

Gene Information

human ... H3C1(8350)
mouse ... H3C1(360198)
rat ... H3C1(679994)

Gene Information

human ... H3F3B(3021)

Gene Information

human ... HIST1H3F(8968)

biological source

rat

biological source

rabbit

biological source

rabbit

biological source

rabbit

isotype

IgG2aκ

isotype

-

isotype

IgG

isotype

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Xiangduo Kong et al.
iScience, 27(4), 109357-109357 (2024-03-21)
Facioscapulohumeral dystrophy (FSHD) is linked to contraction of D4Z4 repeats on chromosome 4q with SMCHD1 mutations acting as a disease modifier. D4Z4 heterochromatin disruption and abnormal upregulation of the transcription factor DUX4, encoded in the D4Z4 repeat, are the hallmarks



Global Trade Item Number

SKUGTIN
MABE243-I04054839168239

Questions

Reviews

No rating value

Active Filters