Skip to Content
Merck
All Photos(1)

Key Documents

ABE44-S

Sigma-Aldrich

Anti-trimethyl Histone H3 (Lys27) Antibody, Trial Size

from rabbit, purified by affinity chromatography

Synonym(s):

H3K27me3, Histone H3 (trimethyl K27), H3 histone family, member T, Histone 3, H3, Histone cluster 3, H3, H3K27me3

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€119.00

€119.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€119.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€119.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

dot blot: suitable
immunocytochemistry: suitable
inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys27)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is a core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Histone H3 lysine 27 methylation (H3K27) is associated with heterochromatin formation and transcriptional repression.

Specificity

Broad species cross-reactivity expected.

Application

Anti-trimethyl Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated on lysine 27 (H3K73me). This purified Ab is specificity verified by dot blot (DB) & validated in WB, ICC, PIA. Small pack size is ideal for screening applications.

Quality

Evaluated by Western Blot in HeLa acid extract and recombinant Histone H3.

Western Blot Analysis: 0.1 µg/ml of this antibody detected histone H3 in 10 µg of HeLa acid extract .

Target description

~ 17 kDa

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danyang Li et al.
International journal of biological sciences, 15(1), 69-80 (2019-01-22)
EZH2 plays vital roles in epigenetic regulation, neuronal development and cancer progression. Here a novel EZH2 variant, namely EZH2-X9 (X9 for short) resulting from alternative splicing, was isolated, identified and functionally characterized. X9 was highly expressed in the brains of
Shumeng Liu et al.
Molecular cell, 67(5), 853-866 (2017-08-15)
Lysine crotonylation (Kcr) is a newly identified histone modification that is associated with active transcription in mammalian cells. Here we report that the chromodomain Y-like transcription corepressor CDYL negatively regulates histone Kcr by acting as a crotonyl-CoA hydratase to convert
Huiming Chen et al.
Oncotarget, 6(15), 13049-13059 (2015-05-07)
Polycomb repressive complex 2 (PRC2), which is responsible for the trimethylation of H3K27 (H3K27me3), plays a part in tumorigenesis, development and/or maintenance of adult tissue specificity. The pivotal role of PRC2 in cancer makes it a therapeutic target for epigenetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service