Skip to Content
Merck

Skip To

LS100

Hood Mounted Pipet Holder, Right Side Mount

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
eCl@ss:
32011202
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Specialty Media

Quality Level

input

sample type cardiac stem cell(s)
sample type induced pluripotent stem cell(s)
sample type: human embryonic stem cell(s)
sample type epithelial cells
sample type mesenchymal stem cell(s)
sample type hematopoietic stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type pancreatic stem cell(s)
sample type neural stem cell(s)

shipped in

ambient

storage temp.

15-25°C

General description

Hood Mounted Pipet Holder Right Side Mount

Application

Research Category
Cell Culture

Packaging

each

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
LS200EP4423000010902888
manufacturer/tradename

Specialty Media

manufacturer/tradename

Specialty Media

manufacturer/tradename

Eppendorf® 4423000010 (Global)

manufacturer/tradename

-

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

-

shipped in

ambient

shipped in

ambient

shipped in

-

shipped in

ambient

input

sample type cardiac stem cell(s)
sample type induced pluripotent stem cell(s)
sample type: human embryonic stem cell(s)
sample type epithelial cells
sample type mesenchymal stem cell(s)
sample type hematopoietic stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type pancreatic stem cell(s)
sample type neural stem cell(s)

input

sample type cardiac stem cell(s)
sample type hematopoietic stem cell(s)
sample type epithelial cells
sample type: mouse embryonic stem cell(s)
sample type mesenchymal stem cell(s)
sample type: human embryonic stem cell(s)
sample type pancreatic stem cell(s)
sample type neural stem cell(s)
sample type induced pluripotent stem cell(s)

input

-

input

-

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

-

storage temp.

no temp limit




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
LS10004053252329012
LS100-DIE-1EA04065267981116

Questions

Reviews

No rating value

Active Filters