Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 mg | Check Cart for Availability | € 874,00 |
About This Item
biological source
rat
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
3G10, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
immunocytochemistry: suitable
isotype
IgG2b
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... EDN1(1906)
Immunogen
Application
Suitable for immunocytochemistry on frozen sections
Optimal working dilutions must be determined by the end user.
Neuroscience
CNS Control of Metabolism
Hormones & Receptors
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Expressed in lung, placental stem villi vessels and in cultured placental vascular smooth muscle cells
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified antibody |
| clone 3G10, monoclonal | clone IC4, monoclonal | clone 3D6, monoclonal | clone TR.ET.48.5, monoclonal |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| biological source rat | biological source mouse | biological source mouse | biological source mouse |
| species reactivity human | species reactivity human | species reactivity human | species reactivity rat, human, sheep |
| Gene Information human ... EDN1(1906) | Gene Information human ... EDN1(1906) | Gene Information human ... EDN1(1906) | Gene Information human ... EDN1(1906) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| CBL87 | 04053252330018 |



