Skip to Content
Merck
All Photos(1)

Key Documents

CBL85

Sigma-Aldrich

Anti-Endothelin-1 Antibody, clone IC4

clone IC4, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€712.00

€712.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€712.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€712.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

IC4, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... EDN1(1906)

Specificity

Recognizes endothelin ET-1 (10% cross-reactivity with ET-3) and when used in conjunction with CBL87 (anti-endothelin C-terminus) has a sensitivity of 0.06 pmol/L for ET-1.

Application

Detect Endothelin-1 using this Anti-Endothelin-1 Antibody, clone IC4 validated for use in IC.
Development of Immunoradiometric assay (IRMA) for endothelin ET-1 which as a circulating plasma peptide has been associated with a variety of disease studies such as diabetes, hypertension, cardiogenic shock, acute myocardial infarction and uraemia.

Suitable for immunohistochemistry on frozen sections

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors

Physical form

Format: Purified
Liquid in 0.02 M PBS with 0.25 M NaCl, pH 7.6. Contains 0.1% sodium azide.
Protein A purified

Storage and Stability

For use within 1 month of purchase when stored at +4°C, for long-term storage, aliquot antibody into small volumes and store at -20°C.

Analysis Note

Control
Expressed in lung, placental stem villi vessels and in cultured placental vascular smooth muscle cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service