Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1.5 mg | Check Cart for Availability | € 198,00 |
About This Item
biological source
rabbit
Quality Segment
conjugate
FITC conjugate
antibody form
F(ab′)2 fragment of affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
mouse
manufacturer/tradename
Chemicon®
technique(s)
immunofluorescence: suitable
shipped in
wet ice
target post-translational modification
unmodified
Application
Secondary & Control Antibodies
Whole Immunoglobulin Secondary Antibodies
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| biological source rabbit | biological source rabbit | biological source rabbit | biological source donkey |
| technique(s) immunofluorescence: suitable | technique(s) immunofluorescence: suitable | technique(s) immunofluorescence: suitable | technique(s) immunofluorescence: suitable |
| species reactivity mouse | species reactivity goat | species reactivity mouse | species reactivity guinea pig |
| conjugate FITC conjugate | conjugate FITC conjugate | conjugate CY3 conjugate | conjugate FITC conjugate |
| antibody form F(ab′)2 fragment of affinity isolated antibody | antibody form F(ab′)2 fragment of affinity isolated antibody | antibody form F(ab′)2 fragment of affinity isolated antibody | antibody form F(ab′)2 fragment of affinity isolated antibody |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 3
Storage Class
11 - Combustible Solids
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



