Skip to Content
Merck
All Photos(1)

Key Documents

AP193F

Sigma-Aldrich

Donkey Anti-Guinea Pig IgG Antibody, FITC conjugate, Species Adsorbed

Chemicon®, from donkey

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€303.00

€303.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
500 μG
€303.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€303.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

donkey

Quality Level

conjugate

FITC conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

guinea pig

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling
This Donkey anti-Guinea Pig IgG Antibody, FITC conjugate, Species Adsorbed is validated for use in IF for the detection of Donkey Guinea Pig IgG.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An appraisal of intermediate filament expression in adult and developing pancreas: vimentin is expressed in alpha cells of rat and mouse embryos.
Di Bella, A; Regoli, M; Nicoletti, C; Ermini, L; Fonzi, L; Bertelli, E
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society null
Rosario Amato et al.
Investigative ophthalmology & visual science, 62(9), 9-9 (2021-07-08)
Glaucoma is a multifactorial disease, causing retinal ganglion cells (RGCs) and optic nerve degeneration. The role of diabetes as a risk factor for glaucoma has been postulated but still not unequivocally demonstrated. The purpose of this study is to clarify
Rosario Amato et al.
Translational vision science & technology, 12(11), 6-6 (2023-11-02)
Glaucoma is an eye-brain axis disorder characterized by loss of retinal ganglion cells (RGCs). Although the role of intraocular pressure (IOP) elevation in glaucoma has been established, the reduction of oxidative stress and inflammation has emerged as a promising target
CXCL14-like immunoreactivity in somatostatin-producing cells of the Japanese quail (Coturnix japonica) pancreas.
Suzuki, et al.
Anatomia, Histologia, Embryologia, 52, 158-162 (2023)
Immunohistochemical localization of monoamine oxidase type B in pancreatic islets of the rat.
Huang, YH; Ito, A; Arai, R
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service