Skip to Content
Merck

Skip To

ABS2249

Anti-SNORC

purified antibody, from rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 μg
Check Cart for Availability
€ 159,00
100 μg
Check Cart for Availability
€ 399,00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Clone:
polyclonal
Species reactivity:
mouse
Application:
ICC, IHC (p), WB
Citations:
-

€ 159,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 12.18 kDa, observed mol wt ~22 kDa

species reactivity

mouse

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, western blot: suitable

isotype

IgG

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Protein SNORC (UniProt: Q9CXL7; also known as Secondary ossification center-associated regulator of chondrocyte maturation protein) is encoded by the Snorc gene (Gene ID: 73234) in murine species. SNORC is a highly conserved, single-pass membrane protein that is synthesized with a signal peptide (aa 1-24), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 25-92), a transmembrane domain (aa 93-113), and a short cytoplasmic domain (aa 114-121). It contains a highly conserved glycosaminoglycan (GAG) attached to serine 44. Its expression is observed only in cartilage and plays a role in the regulation of chondrocyte maturation and postnatal endochondral ossification. It is reported that SNORC is not essential during early differentiation and its function pertains more to mature chondrocytes. (Ref.: Jaiswal, PK., et al. (2020). Sci. Rep. 10; Article 7790).

Immunogen

His-tagged recombinant fragment corresponding to 67 amino acids from the extracellular domain of murine SNORC protein.

Application

Quality Control TestingEvaluated by Western Blotting in lysate from HEK293 cells expressing the untagged Mouse SNORC.Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SNORC in lysate from HEK293 cells expressing the untagged Mouse SNORC.Tested ApplicationWestern Blotting Analysis: A 1:10,000 dilution from a representative lot detected SNORC in 3xFlag mouse-SNORC (Courtesy of Dr. Pierre Moffatt; Shriners Hospitals for Children and McGill University, Montreal, Canada).Immunocytochemistry Analysis: A representative lot detected SNORC in Immunocytochemistry applications (Jaiswal, P.K., et. al. (2020). Sci Rep. 10(1):7790).Immunohistochemistry (Paraffin) Analysis: A 1:3,000 dilution from a representative lot detected SNORC in Mouse Decalcified Femur (Global KO) (Courtesy of Dr. Pierre Moffatt; Shriners Hospitals for Children and McGill University, Montreal, Canada).Immunohistochemistry Applications: A representative lot detected SNORC in Immunohistochemistry applications (Jaiswal, P.K., et. al. (2020). Sci Rep. 10(1):7790).Western Blotting Analysis: A representative lot detected SNORC in Western Blotting applications (Jaiswal, P.K., et. al. (2020). Sci Rep. 10(1):7790).Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-SNORC, Cat. No. ABS2249, is a rabbit polyclonal antibody that detects SNORC and is tested for use in Western Blotting, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Biochem/physiol Actions

This rabbit polyclonal antibody detects secondary ossification center-associated regulator of chondrocyte maturation protein (SNORC). It targets an epitope within 67 amino acids from the extracellular domain.

Physical form

Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide.

Preparation Note

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABN2286ZRB1211ZRB1282
biological source

rabbit

biological source

mouse

biological source

rabbit (recombinant)

biological source

rabbit

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

mol wt

calculated mol wt 12.18 kDa, observed mol wt ~22 kDa

mol wt

calculated mol wt 64.06 kDa, observed mol wt ~58 kDa

mol wt

calculated mol wt 22.61 kDa, observed mol wt ~22 kDa

mol wt

calculated mol wt 17.18 kDa, observed mol wt ~15 kDa

technique(s)

immunocytochemistry: suitable, western blot: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, western blot: suitable

technique(s)

immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, western blot: suitable

technique(s)

affinity binding assay: suitable, immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, western blot: suitable

shipped in

dry ice

shipped in

dry ice

shipped in

ambient

shipped in

ambient


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
ABS2249-25UG04065266020731
ABS2249-100UG04065266020724

Questions

Reviews

No rating value

Active Filters