Skip to Content
Merck
All Photos(11)

Key Documents

ZRB1211

Sigma-Aldrich

Anti-TAGLN Antibody, clone 1D10 , ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

22 kDa actin-binding protein, Protein WS3-10, SM22-alpha, Smooth muscle protein 22-alpha, Transgelin

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€163.00
4 X 25 μL
€336.00

€163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€163.00
4 X 25 μL
€336.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€163.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D10, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 22.61 kDa
observed mol wt ~22 kDa

species reactivity

mouse, human

species reactivity (predicted by homology)

bovine, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TAGLN(6876)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1D10 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Transgelin. It targets an epitope within 12 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 12 amino acids from the C-terminal region of human Transgelin.

Application

Anti-TAGLN, clone 1D10 ZooMAb, Cat. No. ZRB1211, is a highly specific recombinant rabbit monoclonal antibody that detects Transgelin/SM22 alpha and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected TAGLN in HEPG2, NIH-3T3, and human skeletal muscle lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected TAGLN in human uterus tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TAGLN in C2C12 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens and experimental conditions may vary.

Target description

Transgelin (UniProt: Q01995; also known as 22 kDa actin-binding protein, Protein WS3-10, Smooth muscle protein 22-alpha, SM22-alpha) is encoded by the TAGLN (also known as SM22, WS3-10) gene (Gene ID:6876) in human. Transgelin is a shape change sensitive actin-binding protein of the calponin family that contains a calponin-homology domain (aa 24-137) and a C-terminal calponin-like module (aa 175-200) that is essential for actin binding. It is found in vascular and visceral smooth muscles of all adult vertebrates and is considered as an early marker of smooth muscle differentiation. Transgelin expression has also been reported in fibroblasts, spinal cord, adrenal gland, and in several immune cells. It is overexpressed in senescent human fibroblasts. It is shown to be transiently expressed in heart and skeletal muscle cells during early embryogenesis in mouse. Transgelin serves as an actin stress fiber-associated protein that stabilizes actin gels in vitro. It is reported to promote podosome formation and contributes to cell motility in response to injury or inflammation. Activation of TGF 1 signaling is shown to induce Transgelin expression, which enhances mesenchymal stem cells (MSC) differentiation and migration. Over-expression of Transgelin is shown to enhance osteoblast and adipocyte differentiation of MSC and these effects are abolished in the presence of TGF beta signaling inhibitors. Elevated levels of Transgelin are reported to promote tumor metastasis and are correlated with aggressive tumor behavior and poor prognosis in colorectal cancers. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Assinder, SJ et al. (2009). Int. J. Biochem. Cell Biol. 41(3); 482-486; Elsafadi, M., et al. (2016). Cell Death and Disease 7; e2321; Zhou, H-M., et al. (2016). BMC Cancer. 16: 55).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service