Skip to Content
Merck

Skip To

ABE990

Anti-Histone H3.3 K27I

from rabbit

Synonym(s):

H3.3, H3.3 mutant K27I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... H3F3B(3021)

General description

Histone H3.3 (UniProt: P84243) is encoded by the H3F3A (also known as H3.3A, H3F3, PP781, H3F3B, H3.3B) gene (Gene ID: 3020) in human. It is a highly conserved variant form of Histone H3, which replaces conventional H3 in a wide range of nucleosomes in active genes. Histone H 3.3 constitutes the predominant form of histone H3 in non-dividing cells and is incorporated into chromatin independently of DNA synthesis. H3.3 is associated with actively expressed genes in both animals and plants. It is predominantly enriched near transcription end sites (TES) of genes and positively associated with transcription. Histone H3 contains a main globular domain and a long N-terminal tail and is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. For example, acetylation on Lys10 (H3K9ac; initial methionine removed) impairs methylation at Arg9 (H3R8me2s) and acetylation on Lys19 (H3K18ac) and Lys24 (H3K24ac) favors methylation at Arg18 (H3R17me). Phosphorylation at Ser11 (H3S10ph) by Aurora kinase B is crucial for chromosome condensation and cell-cycle progression during mitosis and meiosis. Phosphorylation at Ser11 by RPS6KA4 and RPS6KA5 is important during interphase because it enables the transcription of genes following external stimulation, like mitogens, stress, growth factors or UV irradiation and result in the activation of genes, such as c-Fos and c-Jun. Mutations in Histone H3.3 have been implicated in a high proportion of malignant pediatric brain cancers. The mutant H3.3 histone disrupts epigenetic post-translational modifications near genes involved in cancer processes and in brain function. Histone H3K27 is reported to be methylated by the Polycomb Repressive Complex 2 (PRC2) for X-chromosome inactivation and gene silencing and H3K27M peptides are shown to be potent inhibitors of PRC2 activity. H3K27I peptide also displays inhibitory, although to a lesser extent, on PRC2. The possibility of K27I mutation in Histone H3 has been suggested in pediatric gliomas, which might be due to blockage of glial cell differentiation. (Ref.: Lewis, PW., and Allis, CD (2013). Cell Cycle. 12(20): 3241-3242).
~18 kDa observed; 15.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to 8 amino acids from the N-terminal region of human histone H3.3 with K27I mutation.

Application

Anti-Histone H3.3 K27I, Cat. No. ABE990, is a highly specific rabbit polyclonal antibody that detects Histone H3.3 mutant K27I and has been tested for use in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Histone H3.3 K27I in HEK293 cells transfected with H3.3 K27I mutant, but not the wild-type. (Courtesy of Dr. Peter Lewis, Rockefeller University).

Biochem/physiol Actions

This rabbit polyclonal antibody detects K27I mutant form of histone H3.3 and does not react with the wild-type H3.3.

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in wold-type HEK293 cells and HEK293 cells transfected with Histone H3.3 K27I.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Histone H3.3 K27I in HEK293 cells transfected with Histone H3.3 K27I.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABE1839ABE403MABE872
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rat

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

UniProt accession no.

P84243

UniProt accession no.

P84243

UniProt accession no.

P84243

UniProt accession no.

P84243

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

ChIP: suitable, immunocytochemistry: suitable, western blot: suitable

Gene Information

human ... H3F3B(3021)

Gene Information

human ... H3F3B(3021)

Gene Information

human ... H3F3B(3021)

Gene Information

human ... H3F3B(3021)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
ABE99004054839399701
ABE990-25UL04054839539077

Questions

Reviews

No rating value

Active Filters