Skip to Content
Merck
All Photos(1)

Key Documents

ABE403

Sigma-Aldrich

Anti-Histone H3.3 G34R

from rabbit

Synonym(s):

H3.3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... H3F3B(3021)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
ABE990ABE419MABE940
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

species reactivity

human

species reactivity

human

species reactivity

mouse, human

species reactivity

human

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

ChIP: suitable, immunohistochemistry: suitable, western blot: suitable

technique(s)

ELISA: suitable, dot blot: suitable, immunocytochemistry: suitable, western blot: suitable

shipped in

ambient

shipped in

ambient

shipped in

wet ice

shipped in

wet ice

General description

Histone H3.3 (UniProt: P84243) is encoded by the H3FA3 (also known as H3.3A, H3F3, PP781, H3F3B, H3.3B) gene (Gene ID: 3020) in human. Histones are basic nuclear proteins that are responsible for the nucleosome structure of chromatin in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which DNA is wrapped in repeating units, called nucleosomes, which limits DNA accessibility to the cellular machineries, which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Histone H3.3 is expressed throughout the cell cycle independently of DNA synthesis. It is a variant histone H3 which replaces conventional H3 in a wide range of nucleosomes in active genes. It is the predominant form of histone H3 in non-dividing cells and is incorporated into chromatin independently of DNA synthesis. While H3.3 can function much the same as canonical H3 as a core part of the nucleosome, H3.3 is also deposited into transcriptionally active regions to replace displaced nucleosomes throughout the cell cycle. In tumor cells, H3.3K27M and H3.3G34R/V mutations are heterozygously expressed, with one allele of H3F3A being wildtype. K27M and G34R/V mutations are mutually exclusive in tumors and display distinct gene expression profiles and DNA methylation patterns. Almost all tumors bearing G34R/V mutations also exhibit mutations in ATRX/DAXX and display alternate lengthening of telomeres, which is a classical phenotype of cancerous cells. (Ref.: Yuen, BT., and Knoepfler PS (2013). Cancer cell 24(5); 567-74).

Specificity

This rabbit polyclonal antibody specifically detects G34R mutated form of human Histone H3.3. It does not detect widl-type Histone 3.3.

Immunogen

Conjugated linear peptide correspding to 8 amino acids from the N-terminal region of human histone H3.3 with a G34R mutation.
Epitope: unknown

Application

Anti-Histone H3.3 G34R, Cat. No. ABE403, is a highly specific rabbit polyclonal antibody that targets Histone H3.3 mutant G34R and has been tested for use in Immunohistochemistry and Western Blotting.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in Hek293 cells transfected with Histone 3.3 G34R mutant.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Histone H3.3 G34R in Hek293 cells transfected with Histone 3.3 G34R mutant.

Target description

~17 kDa observed; 15.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service