Skip to Content
Merck

Skip To

ABE76-IM

Anti-Enterobacterio Phage MS2 Coat Protein

from rabbit, purified by affinity chromatography

Synonym(s):

Capsid protein, CP, Coat protein, MS2 Coat Protein, Enterobacterio Phage MS2 Coat Protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 μg
Check Cart for Availability
€ 160,00
100 μg
Check Cart for Availability
€ 388,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
-

€ 160,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB2123

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

virus

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μg

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

General description

The bacteriophage MS2 is a small, male-specific RNA bacteriophage that infects E. coli and other members of the Enterobacteriaceae family. Its genome comprises a single sense (+) strand of RNA 3569 nucleotides in length, which encodes only four proteins: the coat protein, the A or maturation protein, which is required for infectivity, a subunit of the viral replicase, and a small lysis protein encoded in an overlapping reading frame. The capsid protein (UniProt: P03612; also known as CP, Coat protein) of bacteriophage MS2 is a translational repressor. It is reported to inhibit the synthesis of the viral replicase by binding a specific RNA structure that contains the replicase translation initiation region. Capsid protein self-assembles to form an icosahedral capsid with a T=3 symmetry and consists of 89 capsid proteins dimers (178 capsid proteins). It is involved in viral genome encapsidation through the interaction between a capsid protein homodimer and the multiple packaging signals present in the RNA genome. Proline 78 is shown to be essential for viral infectivity. (Ref.: Hill, HR et al. (1997). J. Mol. Biol. 266(1); 1-7).
~14 kDa observed; 13.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

His-tagged recombinant Enterobacteriophage MS2 Coat Protein (aa 2-130).

Application

Anti-EnterobacterioPhage MS2 Coat Protein, Cat. No. ABE76-I, is a highly specific rabbit polyclonal antibody that targets Escherichia phage MS2 (Bacteriophage MS2) Capsid Protein and has been tested in Immunoprecipitation and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A 1:5,000 dilution from a representative lot detected recombinant EnterobacterioPhage MS2 Coat Protein and wildtype MS2 protein (Courtesy of Caroline Horizny from CNSE at SUNY Albany, Albany, NY).

Immunoprecipitation Analysis: A 1:10,000 dilution from a representative lot detected EnterobacterioPhage MS2 Coat Protein in K562 cells treated with Escherichia phage MS2 (Bacteriophage MS2).

Biochem/physiol Actions

This rabbit polyclonal antibody detects MS2 bacteriophage capsid protein. The immunogen used has an N- terminus His tag and mutations to avoid dimerization and capsid formation.

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in K562 treated with Escherichia phage MS2 (Bacteriophage MS2) and K562+MS2 recombinant protein treated with Escherichia phage MS2 (Bacteriophage MS2) Capsid Protein.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected EnterobacterioPhage MS2 Coat Protein in K562 ells treated with Escherichia phage MS2 (Bacteriophage MS2) and K562+MS2 recombinant protein treated with Escherichia phage MS2 (Bacteriophage MS2) Capsid Protein.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MAB131884MABE990MABE284
Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

mouse

biological source

mouse

biological source

mouse

UniProt accession no.

P03612

UniProt accession no.

Q6JC40

UniProt accession no.

Q9HCI7

UniProt accession no.

P43246

clone

polyclonal

clone

7C7, monoclonal

clone

10C3.1, monoclonal

clone

FE11, monoclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters