Skip to Content
Merck
All Photos(4)

Key Documents

MAB131884

Sigma-Aldrich

Anti-BASE-tag Antibody, clone 7C7

clone 7C7, from mouse

Synonym(s):

Capsid protein VP1, Capsid protein AVV9

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€154.00
100 μL
€420.00

€154.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€154.00
100 μL
€420.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€154.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7C7, monoclonal

species reactivity

adenovirus-infected cells, virus

packaging

antibody small pack of 25 μL

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

UniProt accession no.

target post-translational modification

unmodified

General description

Capsid protein VP1 (UniProt: Q6JC40) is encoded by the Cap gene in Adeno-associated virus 9 (AAV9). Adeno-associated viruses (AAV) are small non-enveloped DNA viruses that can serve as promising viral vectors for gene delivery, but can also elicits cellular and humoral immune responses. AAVs are reported to be nonpathogenic in humans and animals and display low immunogenicity. Following their attachment to cell surface receptor they are internalized via endocytosis and their intracellular trafficking is a rate-limiting, slow and inefficient process. AAVs contain an icosahedral capsid, which is 22 to 26 nm in diameter. They are shown to have a single-stranded DNA genome of approximately 4.7 kilobases that is flanked by two T-shaped inverted terminal repeats (ITRs). The 7C7 an epitope-tag antibody recognizes a basic ten amino acid epitope (QPAKKRLNFGQTGD) with high specificity. The basic nature of the BASE-tag epitope sequence distinguishes it from other commonly used epitope tags, such as Myc, EQKLISEEDL, FLAG, DYKDDDDK, HA, YPYDVPDYA, V5 etc. Clone 7C7 recognizes capsid proteins VP1 and VP2, but not VP3. Based on sequence identity of the epitope sequence, this antibody is expected to recognize VP1 and VP2 of AAV serotypes 1,2,3,6,7,8, and 9. (Ref.: Nonnenmacher, M., and Weber, T (2012). Gene Ther. 19(6); 649-658).

Specificity

Clone 7C7 is a mouse monoclonal antibody that detects various serotypes of Adeno-associated virus (AVV). It targets an epitope with in 14 amino acids from the N-terminal region.

Immunogen

Empty Adeno-associated virus 9 (AAV9) Capsids

Application

Anti-BASE-tag, clone 7C7, Cat. No. MAB131884, is a mouse monoclonal antibody that recognizes the epitope QPAKKRLNFGQTGD and has been tested for use in Western Blotting, Immunoprecipitation, and Immunofluorescence.
Immunoprecipitation: A representative lot immunoprecipitated EGFP-BASE-tag-Actin in HEK293T cells transfected with EGFP-BASE-tag-Actin (Courtesy: Dr. Thomas Weber, Icahn School of Medicine at Mount Sinai, New York).
Immunofluorescence: A representative lot detected EGFP-BASE-tag-Actin in HeLa cells transfected with EGFP-BASE-tag-Actin (Courtesy: Dr. Thomas Weber, Icahn School of Medicine at Mount Sinai, New York).
Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting in HEK293 cells transfected with GFP-7C7-Actin.

Western Blotting Analysis: A 1:500 dilution of this antibody detected BASE-tag in HEK293 cells transfected with GFP-7C7-Actin.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service