Skip to Content
Merck

Skip To

ABE476

Anti-GAL4 Antibody (activation domain)

from rabbit, purified by affinity chromatography

Synonym(s):

Regulatory protein GAL4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 428,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
1

€ 428,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB2599

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

yeast

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Saccharomyces cerevisiae ... Gal4(855828)

General description

GAL4 is a transcriptional activator which controls the expression of genes involved in the catabolism of galactose. In addition to a DNA-binding domain, Gal4 contains two acidic regions: activating region I (aa148-238) and activating region II (aa768-881), which can both activate transcription when independently joined to the DNA-binding domain. The activation domains of Gal4 have been shown to interact with Gal80, TBP, SUG1, and ADA2, among other general transcription factors.
~28 kDa, ~38 kDa, ~77 kDa observed

Immunogen

Epitope: C-terminus
GST-tagged recombinant protein corresponding to the C-terminus of yeast GAL4 (activation domain).

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Use Anti-GAL4 (activation domain) Antibody (Rabbit Polyclonal Antibody) validated in WB to detect GAL4 (activation domain) also known as Regulatory protein GAL4.

Biochem/physiol Actions

This antibody recognizes the C-terminus of GAL4 (activation domain).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Gal4 positive and Gal4 negative yeast
Evaluated by Western Blot in Gal4 positive and Gal4 negative yeast cell lysate.

Western Blot Analysis: A 1:160,000 dilution of this antibody detected Gal4 in 5 mg of Gal4 positive (+ recombinant Gal4) yeast cell lysate. This antibody also detected small mounts of endogenous Gal4 in Gal4 negative (- recombinant Gal4) yeast cell lysate.

Other Notes

Replaces: 06-283

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G9293SAB1406938HPA004868
Quality Level

100

Quality Level

200

Quality Level

-

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

technique(s)

western blot: suitable

technique(s)

western blot: 0.5 μg/mL using GAL4-AD fusion protein in Saccharomyces cerevisiae extract

technique(s)

indirect immunofluorescence: suitable, western blot: 1 μg/mL

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunohistochemistry: 1:20-1:50


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
ABE47604053252522741

Questions

Reviews

No rating value

Active Filters