Skip to Content
Merck
All Photos(8)

Key Documents

HPA004868

Sigma-Aldrich

Anti-GALT antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Gal-1-P uridylyltransferase, Anti-UDP-glucose-hexose-1-phosphate uridylyltransferase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Estimated to ship on16 April 2025



Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Estimated to ship on16 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

RANDHQHIRYNPLQDEWVLVSAHRMKRPWQGQVEPQLLKTVPRHDPLNPLCPGAIRANGEVNPQYDSTFLFDNDFPALQPDAPSPGPSDHPLFQAKSARGVCKVMCFHPWSD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GALT(2592)

Looking for similar products? Visit Product Comparison Guide

General description

GALT (galactose-1-phosphate uridylyltransferase) is majorly involved in the Leloir pathway of galactose metabolism. Structural studies have been reported that GALT is a homodimeric molecule. Each monomer consists of six α-helices and a β-sheet formed by nine anti-parallel strands and each monomer contains two active sites for enzymatic activity.

Specificity

Note: The Ensemble Gene ID has changed from ENSG00000137070 in the 46:36 release of the database to ENSG00000213930 in the 48:36 release of the database.

Immunogen

Galactose-1-phosphate uridylyltransferase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GALT (galactose-1-phosphate uridylyltransferase) plays a crucial role in galactose metabolism. It catalyzes the conversion of galactose-1-phosphate to UDP-galactose through a double displacement mechanism. The reaction is highly dependent on the GALT presence. Galactose-1-phosphate (gal-1P) interacts with the uridine diphosphate-glucose (UDP-Glu) to produce uridine diphosphate-galactose (UDP-Gal) and glucose-1-phosphate (glu-1P). GALT facilitates the process of transferring uridine monophosphate (UMP) group to the gal-1P. In humans, alteration in GALT activity causes a classic disease termed as galactosemia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86837

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L J Wong et al.
Biochemistry, 16(5), 1010-1016 (1977-03-08)
Galactose-1-P uridylyltransferase catalyzes the interconversion of UDP-galactose and galactose-1-P with UDP-galactose and glucose-1-P by a double displacement pathway involving a uridylyl-enzyme intermediate. The amount of radioactivity incorporated into the protein by uracil-labeled UDP-glucose is decreased by the presence of UDP-galactose
J E Wedekind et al.
Biochemistry, 34(35), 11049-11061 (1995-09-05)
Galactose-1-phosphate uridylyltransferase catalyzes the reversible transfer of the uridine 5'-monophosphoryl moiety of UDP-glucose to the phosphate group of galactose 1-phosphate to form UDP-galactose. This enzyme participates in the Leloir pathway of galactose metabolism, and its absence is the primary cause
Manshu Tang et al.
Human mutation, 33(7), 1107-1115 (2012-03-31)
Galactose-1-phosphate uridylyltransferase (GALT) catalyzes the conversion of galactose-1-phosphate to UDP-galactose, a key step in the galactose metabolism. Deficiency of GALT activity in humans caused by deleterious variations in the GALT gene can cause a potentially lethal disease called classic galactosemia.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service