Skip to Content
Merck

Skip To

ABE286

Anti-acetyl-p53 (Lys120) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cellular tumor antigen p53, Antigen NY-CO-13, Phosphoprotein p53, Tumor suppressor p53

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
200 μg
Check Cart for Availability
€ 370,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
DB, IP
Citations:
2

€ 370,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), canine (based on 100% sequence homology), rabbit (based on 100% sequence homology), sheep (based on 100% sequence homology)

technique(s)

dot blot: suitable, immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys120)

Gene Information

human ... TP53(7157)

General description

p53 is a DNA-binding, oligomerization domain and transcription activation domain-containing tumor suppressor that upregulates growth arrest and apoptosis-related genes in response to stress signals, thereby influencing programmed cell death, cell differentiation and cell cycle control mechanisms. p53 localizes to the nucleus yet can be chaperoned to the cytoplasm by the negative regulator MDM2, an E3 ubiquitin ligase that is upregulated in the presence of active p53, where MDM2 polyubiquitinates p53 for proteasome targeting. p53 can assemble into tetramers in the absence of DNA, fluctuates between latent and active (DNA-binding) conformations, and is differentially activated through posttranslational modifications including phosphorylation and acetylation. Mutations in the DNA-binding domain (DBD) (amino acids 110-286) of p53 can compromise energetically favorable association with cis elements and are implicated in several human cancers.
~43 kDa calculated. Many isoforms are known to exist. This antibody is predicted to react with isoform 1 (44 kDa), isoform 2 (38 kDa), isoform 3 (39 kDa), isoform 4 (39 kDa), isoform 5 (34 kDa), and isoform 6 (34 kDa).

Immunogen

Epitope: AXIN1 interacting domain
KLH-conjugated linear peptide corresponding to the AXIN1 interacting domain of human p53 acetylated at Lys120.

Application

Anti-acetyl-p53 (Lys120) Antibody is a highly specific rabbit polyclonal antibody, that targets p53 & has been tested in Dot Blot & IP.
Immunoprecipitation Analysis: 1 µg from a representative lot immunoprecipitated acetyl-p53 in camptothecin treated HCT116 cell lysate.

Immunoprecipitation Analysis: 1 µg from a representative lot immunoprecipitated acetyl-p53 in p53 transfected H1299 human lung cotransfected with or without hMOF (Prof. S. McMahon, Kimmel Cancer Center of Thomas Jefferson Medical College).

Immunoprecipitation Analysis: 1.5 µg or 4.5 µg from a representative lot immunoprecipitated endogenous acetyl-p53 in HCT116 cell lysates, which were treated with a vehicle (DMSO) or camptothecin (CPT) (Prof. S. McMahon, Kimmel Cancer Center of Thomas Jefferson Medical College).

Immunoprecipitation Analysis: 1 µg from a representative lot immunoprecipitated acetyl-p53 in H1299 cell lysates, which were transfected with either wild type p53, the 9 K/R mutant (all lysine residues except Lys120 mutated to arginine), the 10 K/R mutant (all lysine residues mutate including Lys 120), or empty vector, with or without hMOF (Prof. S. McMahon, Kimmel Cancer Center of Thomas Jefferson Medical College).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Biochem/physiol Actions

Other homologies: Mouse, Rat, Bovine, Feline, Zebrafish, and Equine (88% sequence homology).
This antibody recognizes the AXIN1 interacting domain of p53 acetylated at Lys120.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Non-acetylated and acetylated p53 peptides
Evaluated by Dot Blot in non-acetylated and acetylated p53 peptides.

Dot Blot Analysis: 1 µg/mL of this antibody detected acetyl-p53 in acetylated p53 peptides

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB9985MABE327SAB4503014
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

clone

polyclonal

clone

polyclonal

clone

DO-1, monoclonal

clone

polyclonal

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

mouse, human, rat


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Batzaya Davaadelger et al.
Oncotarget, 7(19), 27511-27526 (2016-04-07)
The insulin-like growth factor-1 receptor (IGF-1R) signaling pathway is aberrantly activated in multiple cancers and can promote proliferation and chemotherapy resistance. Multiple IGF-1R inhibitors have been developed as potential therapeutics. However, these inhibitors have failed to increase patient survival when
Diana G Wright et al.
Oncotarget, 7(2), 1687-1706 (2015-12-02)
Adult T-cell leukemia (ATL) is an often fatal malignancy caused by infection with the complex retrovirus, human T-cell Leukemia Virus, type 1 (HTLV-1). In ATL patient samples, the tumor suppressor, p53, is infrequently mutated; however, it has been shown to



Global Trade Item Number

SKUGTIN
ABE28604053252861260

Questions

Reviews

No rating value

Active Filters