Skip to Content
Merck
All Photos(2)

Key Documents

AB9985

Sigma-Aldrich

Anti-glutathione p53 (Cys141) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Antigen NY-CO-13, Phosphoprotein p53, Tumor suppressor p53, p53 antigen, p53 transformation suppressor, p53 tumor suppressor, transformation-related protein 53, tumor protein p53

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€316.00

€316.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€316.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€316.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

monkey (100% sequence homology), rat (92% sequence homology), chimpanzee (100% sequence homology), mouse (85% sequence homology), orangutan (100% sequence homology), rabbit (100% sequence homology), ground squirrel (100% sequence homology), rhesus macaque (100% sequence homology), hamster (100% sequence homology), marmoset (100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

glutathionylation (Cys141)

Gene Information

human ... TP53(7157)

General description

Cellular tumor antigen p53 is a member of the p53 family and is found in the cytoplasm, nucleus, and endoplasmic reticulum. It functions as a tumor suppressor within a variety of tumors by either stimulating apoptosis or growth arrest in deference to cell type and physiological factors. p53 serves as a trans-activator, negatively regulating cell division during cell cycle regulation. It is also thought to be involved in the cross-over for Notch signaling. Defects in p53 expression have been implicated in several diseases including; choroid plexus papilloma, lung cancer, head/neck squamous cell carcinomas, esophageal squamous cell carcinoma, Li-Fraumeni syndrome, and hereditary adrenocortival carcinoma. Mutations of the p53 protein have some characteristic features: a) Most of them are missense point mutations giving rise to an altered protein function, and b) Many -but not all- mutant p53 proteins exhibit a common mutant structure, which can be recognized by monoclonal antibodies specific for p53 in the mutant conformation.

Specificity

This antibody recognizes p53 when glutathionylated at Cys141.

Immunogen

Epitope: p53 glutathionylated at Cys141
KLH-conjugated linear peptide corresponding to human p53 glutathionylated at Cys141.

Application

Anti-glutathione p53 (Cys141) Antibody is a rabbit polyclonal antibody for detection of glutathione p53 (Cys141) also known as Antigen NY-CO-13, Phosphoprotein p53, Tumor suppressor p53 & has been validated in WB, IP, IHC, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Western Blot Analysis: A representative lot was used by an independent laboratory in WB. (Dr. Kalkunte S. Srivenugopal, Department of Biomedical Sciences, Texas Tech University Health Sciences Center, 1406 S. Coulter, Amarillo, TX 79106.)

Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in IP. (Yusef, M, et al. (2010). Free Radic Biol Med. 49(5):908-917.)

Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in IC. (Yusef, M, et al. (2010). Free Radic Biol Med. 49(5):908-917.)

Quality

Evaluated by Immunohistochemistry in prostate adenocarcinoma tissue.

Immunohistochemistry Analysis: A 1:500 dilution of this antibody detected glutathione p53 in prostate adenocarcinoma tissue.

Target description

~53 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Prostate adenocarcinoma tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service