Skip to Content
Merck

Skip To

ABC217

Anti-GLI1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Zinc finger protein GLI1, Glioma-associated oncogene, Oncogene GLI

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 340,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
2

€ 340,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1679

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GLI1(2735)

General description

Zinc finger protein GLI1 is also called Glioma-associated oncogene and Oncogene GLI. GLI1 is a transcriptional activator that regulates specific genes during normal development. GLI1 also mediates SHH signaling (and therefore cell proliferation and differentiation) and possibly is involved in craniofacial, central nervous system, gastrointestinal tract, and digital development. GLI1 is expressed in the testis, myometrium, fallopian tube, and brain.
~150 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human GLI1.

Application

Anti-GLI1 Antibody is a highly specific rabbit polyclonal antibody, that targets Gli1 & has been tested in western blotting & IHC (Paraffin).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: A 1:500 dilution from a representative lot detected GL1 in A431, HEK293, HEPG2, and PC3 cells.

Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected GLI1 in human cerebellum and thalamus tissues.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected GLI1 in rat testis tissue.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: A 1:250 dilution of this antibody detected GLI1 in 10 µg of A549 cell lysate.

Other Notes

Replaces: MABC678

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4301901HPA065172SAB2700185
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Natalia Garcia et al.
Reproductive sciences (Thousand Oaks, Calif.), 29(3), 781-790 (2021-10-14)
Uterine leiomyosarcoma (LMS) contributes to a significant proportion of uterine cancer deaths. It is a rare and high-risk gynecological cancer. LMS is challenging to the treatment due to the resistance of several therapies. The activation of the Hedgehog (HH) pathway
Nicolas Jones Villarinho et al.
Cell biology international, 46(9), 1468-1479 (2022-07-12)
Replicative immortality is a key feature of cancer cells and it is maintained by the expression of telomerase, a promising target of novel therapies. Long-term telomerase inhibition can induce resistance, but the mechanisms underlying this process remain unclear. The Sonic



Global Trade Item Number

SKUGTIN
ABC21704053252920646

Questions

Reviews

No rating value

Active Filters