Skip to Content
Merck
All Photos(1)

Key Documents

SAB2700185

Sigma-Aldrich

Anti-Gli1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

AV235269, Gli1, Zfp-5, Zfp5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€444.00

€444.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€444.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€444.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1:500-1:3000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Gli1(14632)

General description

GLI-Kruppel family member Gli1 is a transcriptional activator, which is part of the Gli family of transcription factors. It is a target of hedgehog signaling.

Immunogen

Synthetic peptide containing a sequence corresponding to a region within amino acids 755 and 810 of Gli1 according to NP_034426

Application

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

GLI-Kruppel family member Gli1 may regulate the transcription of specific genes during normal development. It may also play a role in osteoblast formation, craniofacial development and digital development, as well as development of the central nervous system and gastrointestinal tract. The protein mediates SHH signaling and thus cell proliferation and differentiation. Gli1 may also be involved in tissue repair in pancreas. This protein has a protective role during stroke, ischemic injury and Parkinson′s disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The transcription factor GLI1 modulates the inflammatory response during pancreatic tissue remodeling.
Mathew E
The Journal of Biological Chemistry (2014)
Gli1 haploinsufficiency leads to decreased bone mass with an uncoupling of bone metabolism in adult mice.
Kitaura Y
PLoS ONE (2014)
Anusha et al.
Cell death & disease, 12(3), 259-259 (2021-03-13)
Chronic myeloid leukemia (CML) patients with complex chromosomal translocations as well as non-compliant CML patients often demonstrate short-lived responses and poor outcomes on the current therapeutic regimes using Imatinib and its variants. It has been derived so far that leukemic
Gli1 activation and protection against hepatic encephalopathy is suppressed by circulating transforming growth factor ?1 in mice.
McMillin M
Journal of Hepatology (2014)
Fengjiao Liu et al.
CNS neuroscience & therapeutics, 27(12), 1518-1530 (2021-09-05)
Medulloblastoma (MB) is a malignant pediatric brain tumor that arises in the cerebellar granular neurons. Sonic Hedgehog subtype of MB (SHH-MB) is one of the major subtypes of MB in the clinic. However, the molecular mechanisms underlying MB tumorigenesis are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service