Skip to Content
Merck

Skip To

8.55157

MeDbz NovaSyn®TGR resin

Novabiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 159,00
5 g
Check Cart for Availability
€ 624,00

About This Item

NACRES:
NA.22
UNSPSC Code:
13111023

€ 159,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

NovaSyn® TG, Novabiochem®

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

MeDbz NovaSyn TGR resin is a second generation Dbz resin that overcomes the problem of branched peptide formation of the original and provides a robust approach to the synthesis of peptide thioesters by Fmoc SPPS. This product is used exactly in the same manner as standard Dbz resins except that special conditions are not required for loading of the first amino acid. Peptide chain extension is performed on the 4-anilino group, followed by formation of an imidazolidinone (MeNbz) with p-nitrophenyl chloroformate, and cleavage from the resin with TFA. The peptide-MeNbz is used directly in chemical ligation reactions to generate in situ the desired peptide thioester.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.550138.551558.55007
reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

product line

NovaSyn® TG, Novabiochem®

product line

NovaSyn® TG, Novabiochem®

product line

NovaSyn® TG, Novabiochem®

product line

NovaSyn® TG, Novabiochem®

functional group

amine

functional group

amine

functional group

amine

functional group

amine

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-30°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters