Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
product line
NovaSyn® TG, Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
amine
storage temp.
2-30°C
General description
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Literature references
[1] W. Rapp, et al. in ′Innovation & Perspectives in Solid Phase Synthesis, 1st International Symposium′, R. Epton (Eds), SPCC UK Ltd., Birmingham, 1990, pp. 205.
[2] L. Zhang, et al. in ′Peptides 1990, Proc. 21st European Peptide Symposium′, E. Giralt & D. Andreu (Eds), ESCOM, Leiden, 1990, pp.196.
[3] E. Bayer (1991) Angew. Chem. Int. Ed. Engl., 30, 113.
Analysis Note
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.20 - 0.30 mmol/g
Swelling Volume (in CH₂Cl₂): lot specific result
The base resin ist PEG-PS copolymer (90µm), functionalised with an amino group.
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| reaction suitability reaction type: Fmoc solid-phase peptide synthesis | reaction suitability reaction type: Fmoc solid-phase peptide synthesis | reaction suitability reaction type: Fmoc solid-phase peptide synthesis | reaction suitability reaction type: Fmoc solid-phase peptide synthesis |
| product line NovaSyn® TG, Novabiochem® | product line NovaSyn® TG, Novabiochem® | product line NovaSyn® TG, Novabiochem® | product line NovaSyn® TG, Novabiochem® |
| functional group amine | functional group amine | functional group alcohol | functional group amine |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| application(s) peptide synthesis | application(s) peptide synthesis | application(s) peptide synthesis | application(s) peptide synthesis |
| storage temp. 2-30°C | storage temp. 2-30°C | storage temp. 2-30°C | storage temp. 2-8°C |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



