Skip to Content
Merck

Skip To

76174BC

MVP ICON® Buffer Solution, pH 10.0 Blue (Borate-based)

for use with MVP ICON®pH Probe (78088BC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 ea
Check Cart for Availability
€ 25,60

About This Item

UNSPSC Code:
41122409
NACRES:
NA.25

€ 25,60


Check Cart for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

pH calibration

Quality Level

product line

MVP ICON®

form

liquid

manufacturer/tradename

MVP ICON®

application(s)

food and beverages

compatibility

for use with MVP ICON®pH Probe (78088BC)

General description

pH 10.0 Blue Buffer Solution (500 mL) for the MVP ICON® pH ProbeBorate-based buffer solution containing: Patassium carbonate - Potassium Borate - Potassium Hydroxide.

Application

For use in performing 3-point calibration of the MVP ICON® pH/Temperature Probe (78088BC)

Packaging

Clear plastic squeeze bottle with white flip top cap

Legal Information

MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
76173BC76172BC76175BC
product line

MVP ICON®

product line

MVP ICON®

product line

MVP ICON®

product line

MVP ICON®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

form

liquid

form

liquid

form

liquid

form

liquid

manufacturer/tradename

MVP ICON®

manufacturer/tradename

MVP ICON®

manufacturer/tradename

MVP ICON®

manufacturer/tradename

MVP ICON®

description

pH calibration

description

pH calibration

description

pH calibration

description

-


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
7617404065265648745

Questions

Reviews

No rating value

Active Filters