Skip to Content
Merck
All Photos(1)

Key Documents

76173BC

MVP ICON® Buffer Solution, pH 7.0 Yellow

for use with MVP ICON®pH Probe (78088BC)

Sign Into View Organizational & Contract Pricing

Select a Size

76173
€24.10

€24.10


Estimated to ship on26 April 2025


Request more information

Select a Size

Change View
76173
€24.10

About This Item

UNSPSC Code:
41122409

€24.10


Estimated to ship on26 April 2025


Request more information

description

pH calibration

product line

MVP ICON®

form

liquid

manufacturer/tradename

MVP ICON®

application(s)

food and beverages

compatibility

for use with MVP ICON®pH Probe (78088BC)

General description

pH 7.0 Yellow Buffer Solution (500 mL) for the MVP ICON® pH Probe

Application

For use in performing 3-point calibration of the MVP ICON® pH/Temperature Probe (78088BC)

Packaging

Clear plastic squeeze bottle with white flip top cap

Compatibility

Validated for use with MVP ICON® pH/Temperature Probe (78088BC)

Legal Information

MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service