Skip to Content
Merck

Skip To

70976

SpinPrep PCR Clean-up Kit

Rapid purification of PCR products for downstream procedures

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.52
UNSPSC Code:
41105500
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage temp.

10-30°C

General description

Rapid purification of PCR products for downstream procedures
The SpinPrep<TMSYMBOL></TMSYMBOL> PCR Clean-Up Kit is designedfor the rapid purification of DNA amplifiedin PCR. The 10-minute procedure involvesaddition of a binding buffer followed byadsorption of the DNA to a silica membrane ina spin column format. Following a wash step,the DNA is eluted in low-salt buffer. This kitremoves DNA polymerases, dNTPs, salts, and> 99% of primers so that they do not interferewith downstream applications such as cloning,sequencing, or labeling. PCR products from100 bp to > 12,000 bp can be cleaned up, withrecoveries of amplified DNA in the range of60-90% under standard conditions.
Column binding capacity: up to 6 g
PCR volume: 100 l/rxn
Typical recovery: 6090%
Size range: 100 bp to > 12,000 bp
Time required:< 10 min

Other Notes

•82 mlSpinPrep Bind Buffer

•27 mlSpinPrep Wash Buffer

•10 mlSpinPrep Elute Buffer

•100SpinPrep Filters

•100Receiver Tubes

•100Eluate Receiver Tubes

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
70852NA1020UC9601
manufacturer/tradename

Novagen®

manufacturer/tradename

Novagen®

manufacturer/tradename

-

manufacturer/tradename

-

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

-

storage condition

-

storage temp.

10-30°C

storage temp.

10-30°C

storage temp.

15-25°C

storage temp.

15-25°C


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yong H Park et al.
Investigative ophthalmology & visual science, 57(2), 508-526 (2016-02-13)
The α-amino-3-hydroxy-5-methyl-4-isoxazoleproprionic acid (AMPA) receptors (AMPAR) subunits can be posttranscriptionally modified by alternative splicing forming flip and flop isoforms. We determined if an ischemia-like insult to retinal ganglion cells (RGCs) increases AMPAR susceptibility to s-AMPA-mediated excitotoxicity through changes in posttranscriptional



Global Trade Item Number

SKUGTIN
70976-304055977255591

Questions

Reviews

No rating value

Active Filters