Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.52
UNSPSC Code:
41105500
Pricing and availability is not currently available.
manufacturer/tradename
Novagen®
storage condition
OK to freeze
storage temp.
10-30°C
General description
Rapid, efficient extraction of DNA from agarose gels
The SpinPrep<TMSYMBOL></TMSYMBOL> Gel DNA Kit enables efficientextraction of DNA fragments of 150 bp to> 12,000 bp in size from agarose gels. Theprocedure uses GelMelt<TMSYMBOL></TMSYMBOL> Solution to dissolvethe gel slice, followed by adsorption of theDNA to a silica membrane in a spin columnformat. After a wash step, the purified DNAis eluted in low-salt buffer. Each spin columncan bind up to 20 g DNA. Recoveries are consistently in the range of 50-90% understandard conditions. 

Features and Benefits
• No organic extraction or alcohol precipitation
• Total preparation time <30 minutes
• No low melting point agarose is required
• Total preparation time <30 minutes
• No low melting point agarose is required
Other Notes
•5 × 24 mlSpinPrep GelMelt Solution
•27 mlSpinPrep Wash Buffer
•10 mlSpinPrep Elute Buffer
•100SpinPrep Filters
•100Receiver Tubes
•100Eluate Receiver Tubes
•27 mlSpinPrep Wash Buffer
•10 mlSpinPrep Elute Buffer
•100SpinPrep Filters
•100Receiver Tubes
•100Eluate Receiver Tubes
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Multiple Toxicity Values, refer to MSDS (O)
1 of 1
This Item | |||
|---|---|---|---|
| storage temp. 10-30°C | storage temp. 10-30°C | storage temp. - | storage temp. - |
| storage condition OK to freeze | storage condition OK to freeze | storage condition - | storage condition - |
| manufacturer/tradename Novagen® | manufacturer/tradename Novagen® | manufacturer/tradename Cytiva 28-9034-70 | manufacturer/tradename Cytiva 28-9034-71 |
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Liq. 1 - STOT RE 2
target_organs
Thyroid
Storage Class
5.1A - Strongly oxidizing hazardous materials
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





