Skip to Content
Merck

Skip To

573129

STAT3 Inhibitor III, WP1066

InSolution, ≥97%

Synonym(s):

InSolution STAT3 Inhibitor III, WP1066

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C17H14BrN3O
Molecular Weight:
356.22
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥97% (HPLC)
Form:
liquid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light


Quality Segment

assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable AG 490 (Cat. No. 658401) tyrphostin analog that acts as an effective STAT3 pathway inhibitor and a much more potent antitumor agent than AG 490 in inhibiting malignant glioma growth both in cultures in vitro (IC50 = 5.6 and 3.7 µM in a viability assay using U87-MG and U373-MG, respectively; no inhibition against nontumor NHA at 10 µM) and in mice in vivo. Shown to cross blood-brain barrier in mice in vivo.

Packaging

Packaged under inert gas

Physical form

A 50 mM (5 mg/281 µl) solution of STAT3 Inhibitor III, WP1066 (Cat. No. 573097) in DMSO.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
573097573130407625
assay

≥97% (HPLC)

assay

≥97% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

form

liquid

form

solid

form

liquid

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage condition

OK to freeze, protect from light, desiccated (hygroscopic)

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

storage condition

OK to freeze, protect from light


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

188.6 °F - closed cup - (refers to pure substance)

flash_point_c

87 °C - closed cup - (refers to pure substance)



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

The Janus kinase (JAK)/signal transducers and activators of transcription (STAT) signaling pathway plays an important role in cell proliferation, cell differentiation, cell migration, and cell death. It is the principal signaling mechanism for a variety of cytokines and growth factors. Constitutive activation or dysregulation of JAK/STAT signaling can result in inflammatory disease, erythrocytosis, gigantism, and leukemia.






Global Trade Item Number

SKUGTIN
573129-5MG04055977190199

Questions

Reviews

No rating value

Active Filters