Skip to Content
Merck
All Photos(2)

Key Documents

407625

Sigma-Aldrich

STAT3 Inhibitor XVI

The STAT3 Inhibitor XVI controls the biological activity of STAT3. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

STAT3 Inhibitor XVI, IL-6 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€250.00

€250.00


Estimated to ship on29 May 2025



Select a Size

Change View
5 MG
€250.00

About This Item

Empirical Formula (Hill Notation):
C21H22N2O3S3
CAS Number:
Molecular Weight:
446.61
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€250.00


Estimated to ship on29 May 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

gray

solubility

DMSO: 50 mg/mL, light pink-purple

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NCC1=CC=CS1)C2=CC(S(N3CCCC3)(=O)=O)=C(C4=CC=C(C)C=C4)S2

InChI

1S/C21H22N2O3S3/c1-15-6-8-16(9-7-15)20-19(29(25,26)23-10-2-3-11-23)13-18(28-20)21(24)22-14-17-5-4-12-27-17/h4-9,12-13H,2-3,10-11,14H2,1H3,(H,22,24)

InChI key

NVANVFNPZWSWRM-UHFFFAOYSA-N

General description

A cell-permeable, arylsulphonamidyl thiophene amide that is shown to reduce IL-6-induced STAT3 transcriptional activity (EC50 = 15 µM) in a luciferase reporter assay, in HeLa cells. It selectively inhibits STAT3, but not STAT1, phosphorylation following IL-6 stimulation in both HeLa and MDA-MB-231 breast cancer cells at concentrations of 10-30 &micro:M and 30-100 µM, respectively. Following IL-6 stimulation, it suppresses the cell viability of STAT3-dependent cancer cells by 40% compared with that of STAT3-null cancer cells at the low micromolar level.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zinzalla, G., et al. 2010. Bioorg Med Chem Lett20, 7029.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service