Skip to Content
Merck

Skip To

239803

Cyclopamine

from Veratrum californicum, ≥97% (HPLC), solid, Sonic Hedgehog signaling antagonist, Calbiochem®

Synonym(s):

Cyclopamine, V. californicum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 291,00
5 mg
Check Cart for Availability
€ 449,00

About This Item

Empirical Formula (Hill Notation):
C27H41NO2
CAS Number:
Molecular Weight:
411.62
UNSPSC Code:
12352116
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light

€ 291,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Cyclopamine, V. californicum, Cyclopamine, V. californicum, CAS 4449-51-8, is a cell-permeable steroidal alkaloid & cholesterol mimic that specifically antagonizes Sonic Hedgehog signaling through direct interaction with Smo.

Quality Segment

description

RTECS - GY0750000

assay

≥95% (HPLC)

form

solid

potency

20 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMF: 10 mg/mL, DMSO: 4 mg/mL, ethanol: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

N1[C@@H]2[C@H](O[C@@]3([C@@H]2C)CC[C@H]4[C@H]5[C@@H]([C@]6(CC[C@@H](CC6=CC5)O)C)CC4=C3C)C[C@@H](C1)C

InChI

1S/C27H41NO2/c1-15-11-24-25(28-14-15)17(3)27(30-24)10-8-20-21-6-5-18-12-19(29)7-9-26(18,4)23(21)13-22(20)16(27)2/h5,15,17,19-21,23-25,28-29H,6-14H2,1-4H3/t15-,17+,19-,20-,21-,23-,24+,25-,26-,27-/m0/s1

InChI key

QASFUMOKHFSJGL-LAFRSMQTSA-N

General description

A cell-permeable steroidal alkaloid and cholesterol mimic that displays both teratogenic and antitumor activities. It disrupts cholesterol bio-synthesis and specifically antagonizes shh (Sonic Hedgehog) signaling pathway through direct interaction with Smo (smoothened), a distant relative of G-protein-coupled receptors. A valuable tool for studying the involvement of shh signaling in the development of various tumors. Tomatidine can be used as a negative control. Also available as a 10 mM solution in Ethanol (Cat. No. 239806).
Cyclopamine, V. californicum, CAS 4449-51-8, is a cell-permeable steroidal alkaloid & cholesterol mimic that specifically antagonizes Sonic Hedgehog signaling through direct interaction with Smo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Hh signaling in Shh-light2 assay
Product does not compete with ATP.

Preparation Note

Ethanol stock solutions may require warming to achieve complete solubilization.
Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Berman, D.M., et al. 2003. Nature,425, 846.
Thayer, S.P., et al. 2003. Nature,425, 851.
Watkins, D.N., et al. 2003. Nature422, 313.
Chen, J.K., et al. 2002. Proc. Natl. Acad. Sci. USA99, 14071.
Berman, D.M., et al. 2002. Science297, 1559.
Taipale, J., et al. 2000. Nature406, 1005.
Kim, S.K., and Melton, D.A. 1998. Proc. Natl. Acad. Sci. USA95, 13036.
Cooper, M.K., et al. 1998. Science280, 1603.
Incardona, J.P., et al. 1998. Development125, 3553.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Carcinogenic / Teratogenic (D)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
239806239804234115
form

solid

form

liquid

form

solid

form

powder

assay

≥97% (HPLC)

assay

≥97% (NMR)

assay

≥70% (sum of two isomers, HPLC)

assay

≥94% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage temp.

−20°C

storage temp.

−70°C

storage temp.

−20°C

storage temp.

10-30°C

solubility

DMF: 10 mg/mL, ethanol: 5 mg/mL, DMSO: 4 mg/mL

solubility

-

solubility

ethanol: 1 mg/mL, methanol: 1 mg/mL, DMSO: 5 mg/mL

solubility

ethanol: 10 mg/mL, water: soluble


Still not finding the right product?

Explore all of our products under Cyclopamine, V. californicum


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
239803-1MG04055977198867

Questions

Reviews

No rating value

Active Filters