Skip to Content
Merck

Skip To

239804

Cyclopamine-KAAD

≥70% (sum of two isomers, HPLC), solid, Hedgehog signaling inhibitor, Calbiochem®

Synonym(s):

Cyclopamine-KAAD, 3- Keto-N-( aminoethyl- aminocaproyl- dihydrocinnamoyl)cyclopamine, KAAD-Cyclopamine, Shh Signaling Antagonist II, 3-Keto-N-(aminoethyl-aminocaproyl-dihydrocinnamoyl)cyclopamine, KAAD-Cyclopamine, Shh Signaling Antagonist II

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 292,00
500 μg
Check Cart for Availability
€ 1.060,00

About This Item

Empirical Formula (Hill Notation):
C44H63N3O4
Molecular Weight:
697.99
UNSPSC Code:
12352116
NACRES:
NA.77
Assay:
≥70% (sum of two isomers, HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light

€ 292,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Cyclopamine-KAAD, Cyclopamine- KAAD, CAS 306387-90-6, is a cell-permeable potent analog of Cyclopamine (Cat. No. 239803) that specifically inhibits Hedgehog (Hh) signaling with similar or lower toxicity (IC50 = 20 nM).

Quality Segment

assay

≥70% (sum of two isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white to light yellow

solubility

ethanol: 1 mg/mL, methanol: 1 mg/mL, DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A potent, cell-permeable analog of Cyclopamine (Cat. No. 239803) that specifically inhibits the Hedgehog (Hh) signaling with similar or lower toxicity (IC50 = 20 nM in Shh-LIGHT2 assay; 50 nM in p2Ptch-/-cells; 500 nM in SmoA1-LIGHT cells). Binds to SmoA1 and promotes its exit from the endoplasmic reticulum. Suppresses both the ShhNp-induced pathway activity and SmoA1-induced reporter activity. Shown to sensitize human glioma cells to TRAIL-induced apoptosis. Also available as a 1 mM solution in DMSO (Cat. No. 239807).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Hh signaling in Shh-light2 assay
Product does not compete with ATP.
Reversible: no
Target IC50: 20 nM against Hedgehog (Hh) signaling in Shh-LIGHT2 assay; 50 nM in p2Ptch-/-cells; 500 nM in SmoA1-LIGHT cells

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 weeks at -20°C.

Other Notes

Siegelin, M.D. et al. 2009. Neurobiol. Dis.34, 259.
Watkins, D.N., et al. 2003. Nature422, 313.
Berman, D.M., et al. 2002. Science297, 1559.
Chen, J.K., et al. 2002. Proc. Natl. Acad. Sci. USA99, 14071.
Chen, J.K., et al. 2002. Genes Dev.16, 2743.
Frank-Kamenetsky, M., et al. 2002. J. Biol.1, 10.
Taipale, J., et al. 2000. Nature406, 1005.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
239807239803239806
assay

≥70% (sum of two isomers, HPLC)

assay

≥70% (sum of two isomers, HPLC)

assay

≥97% (HPLC)

assay

≥97% (NMR)

form

solid

form

liquid

form

solid

form

liquid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−70°C

storage temp.

−20°C

storage temp.

−70°C

solubility

ethanol: 1 mg/mL, DMSO: 5 mg/mL, methanol: 1 mg/mL

solubility

-

solubility

DMF: 10 mg/mL, DMSO: 4 mg/mL, ethanol: 5 mg/mL

solubility

-


Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters