Skip to Content
Merck

Skip To

239779

CHES

ULTROL® Grade, ≥99% (titration), zwitterionic buffer useful in pH range 8.6-10.0

Synonym(s):

CHES, ULTROL® Grade, 2-(N-Cyclohexylamino)ethanesulfonic Acid

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12161700
NACRES:
NA.25
MDL number:


Quality Segment

assay

≥99% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 M

cation traces

heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])CCNC1CCCCC1

InChI

1S/C8H17NO3S.Na/c10-13(11,12)7-6-9-8-4-2-1-3-5-8;/h8-9H,1-7H2,(H,10,11,12);/q;+1/p-1

InChI key

TXWKJOVDGFRVOG-UHFFFAOYSA-M

General description

(Absorbance 290nm, 1.0M, ≤0.05)
A zwitterionic buffer that is useful in the pH 8.6-10.0 range.
Biological buffer (Absorbance 290nm, 1.0M, ≤0.05)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
239782391340475898
assay

≥99% (titration)

assay

≥99% (HPLC)

assay

≥99% (alkalimetric)

assay

≥99% (titration)

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

form

solid

form

solid

form

solid

form

solid

solubility

water: 1 M

solubility

water: 20 mg/mL

solubility

water: 400 mg/mL, aqueous base: soluble

solubility

water: 1 M

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
239779-100GM07790788048921

Questions

Reviews

No rating value

Active Filters