Skip to Content
Merck
All Photos(1)

Key Documents

239782

Millipore

CAPS

ULTROL® Grade, ≥99% (HPLC), zwitterionic buffer useful in pH range 7.9-11.1

Synonym(s):

CAPS, ULTROL® Grade, 3-(Cyclohexylamino)propanesulfonic Acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
Inquire


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
Inquire

About This Item

Empirical Formula (Hill Notation):
C9H19NO3S
CAS Number:
Molecular Weight:
221.32
MDL number:
UNSPSC Code:
12161703
NACRES:
NA.25


Estimated to ship on19 May 2025


Request a Bulk Order

description

Merck USA index - 14, 1767

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pKa (25 °C)

10.0-10.6

solubility

water: 20 mg/mL

cation traces

heavy metals: ≤1 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

[S](=O)(=O)([O-])CCC[N+H2]C1CCCCC1

InChI

1S/C9H19NO3S/c11-14(12,13)8-4-7-10-9-5-2-1-3-6-9/h9-10H,1-8H2,(H,11,12,13)

InChI key

PJWWRFATQTVXHA-UHFFFAOYSA-N

General description

A zwitterionic buffer that is useful in the pH 7.9-11.1 range. Has a pKa of 10.0-10.6 at 25°C.
Absorbance (100 mM, 260 nm):≤0.05.
Biological buffer (absorbance: <0.05 at 260 nm, 100 mM; pKa: 10.0-10.6 at 25°C).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 6 months at room temperature.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service