Skip to Content
Merck

Skip To

233152

Dithioerythritol

A protective agent for SH groups.

Synonym(s):

Dithioerythritol, Cleland’s Other Reagent, DTE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Check Cart for Availability
Inquire

About This Item

Empirical Formula (Hill Notation):
C4H10O2S2
CAS Number:
Molecular Weight:
154.25
UNSPSC Code:
12161504
NACRES:
NA.25
MDL number:

Inquire

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

RTECS - KF2410000

assay

≥98% (titration)

form

solid

reaction suitability

reagent type: reductant

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1% disulfides (UV)

color

white

pH

4.5-6.5 (25 °C, 15.4 g/L)

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

SC[C@H](O)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4+

InChI key

VHJLVAABSRFDPM-ZXZARUISSA-N

General description

A protective agent for SH groups.
Water-soluble, cell-permeable protective reagent for sulfhydryl groups. Maintains monothiols in the reduced state and reduces disulfides quantitatively. Protects protein sulfhydryls and restores enzyme activity lost by the oxidation of sulfhydryl groups in vitro.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C) under an inert gas. Stock solutions are stable for up to 2 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D0632D9779233155
assay

≥98% (titration)

assay

≥98% (HPLC), ≥99.0% (titration)

assay

≥98% (HPLC), ≥99% (titration)

assay

≥97% (titration)

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

solubility

water: 50 mg/mL

solubility

H2O: soluble 50 mg/mL, clear, colorless to very faintly yellow

solubility

H2O: 50 mg/mL

solubility

water: 30 mg/mL, ethanol: soluble

impurities

<1% disulfides (UV)

impurities

-

impurities

-

impurities

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

form

solid

form

powder

form

powder

form

powder


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters