Skip to Content
Merck

Skip To

233155

Cleland′s Reagent

A protective agent for SH groups. Cyclizes as it reduces disulfides to thiols, so reaction is "driven" to completion. Does not contain waxy surfactant found in many other preparations. Oxidized dithiothreitol: ≤0.5%.

Synonym(s):

Cleland’s Reagent, DTT, Dithiothreitol, Dithiothreitol, DTT

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C4H10O2S2
CAS Number:
Molecular Weight:
154.25
UNSPSC Code:
12161504
NACRES:
NA.25
MDL number:
Assay:
≥97% (titration)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

Merck USA index - 14, 3376

assay

≥97% (titration)

form

powder

reaction suitability

reagent type: reductant

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

water: 30 mg/mL, ethanol: soluble

cation traces

heavy metals: ≤1 ppm, heavy metals: <1 ppm

shipped in

ambient

storage temp.

2-8°C

SMILES string

SC[C@H](O)[C@@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m0/s1

InChI key

VHJLVAABSRFDPM-IMJSIDKUSA-N

General description

A protective agent for SH groups. Cyclizes as it reduces disulfides to thiols, so reaction is "driven" to completion. Does not contain waxy surfactant found in many other preparations.
DTT is a water-soluble protective reagent for sulfhydryl groups. It maintains monothiols in the reduced state and reduces disulfides quantitatively. Most widely used for the study of disulfide exchange reactions of protein disulfides. DTT readily permeates cell membranes. Protects protein sulfhydryls and restores enzyme activity lost by the oxidation of sulfhydryl groups in vitro. As a reducing agent, it cyclizes as it reduces disulfides to thiols, so the reaction is "driven" to completion without using large excesses of reductant. If free thiols are to be assayed later, excess reductant must be removed by dialysis or some other appropriate method. Alternatively, an immobilized form of DTT (REDUCTACRYL Reagent, Cat. No. 233157) can be used. Oxidized dithiothreitol: ≤0.5%.
Oxidized dithiothreitol: ≤0.5%.

Application


  • Cyclic nucleotide phosphodiesterase in Dictyostelium discoideum: interconversion of two enzyme forms.: Discusses the role of Cleland′s Reagent in differentiating forms of cyclic nucleotide phosphodiesterase, showcasing its utility in enzymatic studies and its broader implications in biochemistry research (Chassy BM, 1972).

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot, purge with inert gas, and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gailis, L. 1994. Eur. J. Pharmacol. 292, 39.
Lauriault, V.V.M., and O′Brien, P.J. 1991. Mol. Pharmacol.40, 125.
Klonne, D.R., and Johnson, D.R. 1988. Toxicol. Lett.42, 199.
Zahler, W.L., and Cleland, W.W. 1968. J. Biol. Chem.243, 716.
Cleland, W.W. 1964. Biochemistry3, 480.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
233153-M233156560000
assay

≥97% (titration)

assay

≥99% (titration)

assay

≥99% (titration)

assay

-

solubility

water: 30 mg/mL, ethanol: soluble

solubility

water: 200 mM, ethanol: soluble

solubility

water: 15 mg/mL

solubility

-

reaction suitability

reagent type: reductant

reaction suitability

reagent type: reductant

reaction suitability

reagent type: reductant

reaction suitability

reagent type: reductant

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

15-25°C

form

powder

form

solid

form

powder, solid

form

liquid


Still not finding the right product?

Explore all of our products under Cleland’s Reagent


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters