Skip to Content
Merck

Skip To

219670

CFTR Inhibitor-172

≥95% (HPLC), solid, CFTR inhibitor, Calbiochem®

Synonym(s):

CFTR Inhibitor-172, 3-[(3-Trifluoromethyl)phenyl]-5-[(4-carboxyphenyl)methylene]-2-thioxo-4-thiazolidinone, CFTR inh-172, 3-[(3-Trifluoromethyl)phenyl]-5-[(4-carboxyphenyl)methylene]-2-thioxo-4-thiazolidinone, CFTRinh-172

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 184,00

About This Item

Empirical Formula (Hill Notation):
C18H10F3NO3S2
CAS Number:
Molecular Weight:
409.40
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light

€ 184,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

CFTR Inhibitor-172, CFTR Inhibitor-172, CAS 307510-92-5, is a cell-permeable, potent, reversible, rapid, and voltage-independent inhibitor of CFTR-mediated Cl- transport in human airway cells (Ki ~ 300 nM).

Quality Segment

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(ccc1)N2C(=S)S\C(=C/c3ccc(cc3)C(=O)O)\C2=O

InChI

1S/C18H10F3NO3S2/c19-18(20,21)12-2-1-3-13(9-12)22-15(23)14(27-17(22)26)8-10-4-6-11(7-5-10)16(24)25/h1-9H,(H,24,25)/b14-8-

InChI key

JIMHYXZZCWVCMI-ZSOIEALJSA-N

General description

A cell-permeable 2-thio-4-thiazolidinone compound that acts as a potent, reversible, rapid, and voltage-independent inhibitor of CFTR (cystic fibrosis transmembrane conductance regulator)-mediated Cl- transport in human airway cells (Ki ~ 300 nM). It does not prevent elevation of cellular cAMP, nor does it inhibit non-CFTR Cl- channels, MDR-1, ATP-sensitive K+ channels, or a series of other transporters even at concentrations as high as 5 µM. Shown to block cholera toxin-induced intestinal fluid secretion in mice. May be also useful for reducing intestinal fluid loss in other secretory diarrheas. Also available as a 50 mM solution in DMSO (Cat. No. 219674).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CFTR
Product does not compete with ATP.
Reversible: yes
Target Ki: ~ 300 nM against CFTR (cystic fibrosis transmembrane conductance regulator)-mediated Cl- transport in human airway cells

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yang, H., et al. 2003. J. Biol. Chem.278, 35079.
Al-Awqati, Q., et al. 2002. J. Clin. Invest.110, 1599.
Ma, T., et al. 2002. J. Clin. Invest.110, 1651.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C2992220486528108
assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

form

solid

form

powder

form

solid

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 5 mg/mL

solubility

DMSO: ≥10 mg/mL, H2O: insoluble

solubility

DMSO: 5 mg/mL

solubility

DMSO: 10 mg/mL, ethanol: 5 mg/mL


Still not finding the right product?

Explore all of our products under CFTR Inhibitor-172


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters