Skip to Content
Merck

Skip To

220486

Chk2 Inhibitor II

≥95% (HPLC), Chk2 inhibitor, solid

Synonym(s):

Chk2 Inhibitor II, 2-(4-(4-Chlorophenoxy)phenyl)-1H-benzimidazole-5-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 213,00

About This Item

Empirical Formula (Hill Notation):
C20H14ClN3O2
CAS Number:
Molecular Weight:
363.80
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light

€ 213,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Chk2 Inhibitor II, Chk2 Inhibitor II, BML-277, CAS 516480-79-8 is a cell-permeable, potent, reversible, ATP-competitive inhibitor of Chk2 (IC50 = 15 nM).

Quality Segment

assay

≥95% (HPLC)

form

solid

potency

15 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

pale yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)Oc2ccc(cc2)c3[nH]c4c(n3)cc(cc4)C(=O)N

InChI

1S/C20H14ClN3O2/c21-14-4-8-16(9-5-14)26-15-6-1-12(2-7-15)20-23-17-10-3-13(19(22)25)11-18(17)24-20/h1-11H,(H2,22,25)(H,23,24)

InChI key

UXGJAOIJSROTTN-UHFFFAOYSA-N

General description

A cell-permeable and reversible benzimidazolo compound that acts as a potent and ATP-competitive inhibitor of Chk2 with an IC50 of 15 nM and a Ki of 37 nM. Displays ~ 1,000-fold greater selectivity over Cdk1/B and CK1 (IC50 = 12 M and 17 M, respectively) and only weakly affects the activities of a panel of 31 kinases (<25% inhibition at 10 M), including Chk1. Shown to rescue both peripheral CD4+ and CD8+ T-cells from γ-irradiation-induced apoptosis with an EC50 of 3 M and 7.6 M, respectively. Also available as a 25 mM solution in DMSO (Cat. No. 220491).

Biochem/physiol Actions

Cell permeable: yes
Ki = 37 nM inhibiting Chk2.EC50 = 3 µM and 7.6 µM in rescuing both peripheral CD4+ and CD8+ T-cells from γ-irradiation-induced apoptosis
Primary Target
Chk2
Product competes with ATP.
Reversible: yes
Target Ki: 37 nM inhibiting Chk2

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Arienti, K. L., et al. 2005. J. Med. Chem.48, 1873.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
238811220488218860
assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥99% (HPLC)

form

solid

form

solid

form

solid

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

storage condition

OK to freeze, protect from light


Still not finding the right product?

Explore all of our products under Chk2 Inhibitor II


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters