Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 25 mg | Check Cart for Availability | € 324,00 |
About This Item
Product Name
CENTA β-Lactamase Substrate, CENTA β-Lactamase Substrate is a chromogenic β-lactamase substrate for both Gram-negative and Gram-positive bacteria.
Quality Segment
description
RTECS - OD2265000
assay
≥98% (TLC)
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
yellow
solubility
25 mM phosphate buffer, pH 7.0: 2.5 mg/mL, methanol: 50 mg/mL
shipped in
ambient
storage temp.
−20°C
General description
Please note that the molecular weight for this compound is batch-specific due to variable water content.
Preparation Note
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98% (TLC) | assay - | assay - | assay - |
| form powder | form lyophilized powder | form lyophilized | form lyophilized powder |
| Quality Level 100 | Quality Level 200 | Quality Level 100 | Quality Level 300 |
| manufacturer/tradename Calbiochem® | manufacturer/tradename - | manufacturer/tradename Calbiochem® | manufacturer/tradename - |
| storage temp. −20°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C |
| solubility 25 mM phosphate buffer, pH 7.0: 2.5 mg/mL, methanol: 50 mg/mL | solubility - | solubility sterile distilled water: soluble | solubility - |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 219475-25MG | 04055977218527 |



