Skip to Content
Merck
All Photos(2)

Key Documents

P4524

Sigma-Aldrich

β-Lactamase from Enterobacter cloacae

Type IV, lyophilized powder, 0.2-0.6 units/mg protein (using benzylpenicillin)

Synonym(s):

β-Lactamase I, β-Lactamase II, Cephalosporinase, Penicillin amido-β-lactam hydrolase, Penicillinase from Enterobacter cloacae

Sign Into View Organizational & Contract Pricing

Select a Size

100 UNITS
€306.00
250 UNITS
€614.00

€306.00


Check Cart for Availability


Select a Size

Change View
100 UNITS
€306.00
250 UNITS
€614.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

€306.00


Check Cart for Availability

biological source

bacterial (Enterobacter cloacae)

Quality Level

type

Type IV

form

lyophilized powder

specific activity

0.2-0.6 units/mg protein (using benzylpenicillin)

mol wt

29 kDa

composition

Protein, ~60%

technique(s)

enzyme immunoassay: suitable

suitability

suitable for enzyme test

application(s)

pharmaceutical

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

β-Lactamase is a monomeric enzyme of 29 kDa.[1] This product is produced from Enterobacter cloacae and is provided as a lyophilized powder. It acts as a target protein for β-lactam antibiotics. This enzyme is present in Gram-negative bacteria.[2]

Application

β--lactamase is used to inactivate β-lactam antibiotics by breaking open the β-lactam ring. β--lactamase is used to study antibiotic resistance and resistance suppression. Product P4524 is produced from Enterobacter cloacae and is provided as a lyophilized powder.
β-Lactamase from Enterobacter cloacae has been used in the maternal and fetal quantitative blood cultures to avoid any carryover phenomenon. It has also been used as a component in Mueller Hinton (MH) broth for placental cultures.[3]

Biochem/physiol Actions

β--lactamase inactivates β-lactam antibiotics by breaking open the β-lactam ring.

Unit Definition

One unit will hydrolyze 1.0 μmole of benzylpenicillin per min at pH 7.0 at 25 °C. This International Unit (using benzylpenicillin as substrate) is approximately equal to 600 Levy or 75 Pollock units.

Physical form

Lyophilized powder containing sodium phosphate buffer salts

Analysis Note

Protein determined by biuret.

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Persistent bacteremia in rabbit fetuses despite maternal antibiotic therapy in a novel intrauterine-infection model
Gras-Le Guen C, et al.
Antimicrobial Agents and Chemotherapy, 47(7), 2125-2130 (2003)
Protein-protein interactions monitored in mammalian cells via complementation of beta-lactamase enzyme fragments
Wehrman T, et al.
Proceedings of the National Academy of Sciences of the USA, 99(6), 3469-3474 (2002)
Past and present perspectives on beta$-lactamases
Bush K
Antimicrobial Agents and Chemotherapy, 62(10) (2018)
P J Johnsen et al.
Genetics, 181(4), 1521-1533 (2009-02-05)
We present a new hypothesis for the selective pressures responsible for maintaining natural competence and transformation. Our hypothesis is based in part on the observation that in Bacillus subtilis, where transformation is widespread, competence is associated with periods of nongrowth
Jung-Ho Shin et al.
EMBO reports, 22(2), e51790-e51790 (2021-01-20)
Bactericidal antibiotics are powerful agents due to their ability to convert essential bacterial functions into lethal processes. However, many important bacterial pathogens are remarkably tolerant against bactericidal antibiotics due to inducible damage repair responses. The cell wall damage response two-component

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service