Skip to Content
Merck

Skip To

17-348

Dimethyl Arginine Antibody Sampler Pack

Upstate®, from rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12161503
NACRES:
NA.41
eCl@ss:
32161000
Species reactivity:
human, mouse
Technique(s):
immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
Application:
ICC, IHC, IP, WB
Citations:
7
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

shipped in

wet ice

target post-translational modification

dimethylation (Arg)

Application

Dimethyl Arginine Antibody Sampler Pack. Routinely evaluated by western blot on Jurkat Cell Lysate.

Packaging

Kit capacity: Depends upon application

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.
Routinely evaluated by western blot on Jurkat Cell Lysate

Other Notes

Anti-dimethyl-Arginine, asymmetric

Anti-dimethyl-Arginine, symmetric

Anti-dimethyl-Arginine, symmetric

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
07-41307-41209-814
species reactivity

human, mouse

species reactivity

human, mouse

species reactivity

mouse, human

species reactivity

mouse

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, immunohistochemistry: suitable, western blot: suitable

technique(s)

western blot: suitable

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

western blot: suitable

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class

10 - Combustible liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters