Skip to Content
Merck
All Photos(1)

Key Documents

09-814

Sigma-Aldrich

Anti-dimethyl-Arginine Antibody, asymmetric (ASYM25)

serum, from rabbit

Synonym(s):

dimethyl-arginine, asymmetric

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€509.00

€509.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€509.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€509.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology), human (based on 100% sequence homology)

technique(s)

western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Asymmetrical dimethlarginine (ADMA) is an endogenous, L-arginine metabolite arising from the proteolysis of methylated arginine containing proteins and is a known inhibitor of nitric oxide synthase (NOS). In various studies, ADMA has been observed as having a regulatory role in cell motility and the actin cytoskeleton. Elevated levels of ADMA are associated with sporadic FSGS, coronary artery disease, hypertension, peripheral arterial occlusive disease, obesity, stroke, preeclampsia, and cardiovascular mortality/morbidity in patients with diabetic nephropathy.

Specificity

This antibody recognizes dimethylated arginines.

Immunogen

KLH-conjugated linear peptide containing asymmetric dimethyl-arginine-glycine repeats.

Application

Detect dimethyl-Arginine using this Anti-dimethyl-Arginine Antibody, asymmetric (ASYM25) validated for use in WB.

Quality

Evaluated by Western Blot in PRMT1 treated and untreated MEF cell lysates.

Western Blot Analysis: 1:1,000 dilution of this antibody detected dimethyl-Arginine on 10 µg of PRMT1 treated and untreated MEF cell lysates.

Target description

Multiple. This antibody detects proteins containing asymmetrical dimethylated arginines.

Analysis Note

Control
PRMT1 treated and untreated MEF cell lysates

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Ming Liu et al.
Cancer research, 79(11), 2865-2877 (2019-04-25)
C/EBPα is an essential transcription factor involved in regulating the expression or function of certain cell-cycle regulators, including in breast cancer cells. Although protein arginine methyltransferases have been shown to play oncogenic roles in a variety of cancers, little is
Yongtai Bai et al.
Molecular cell, 75(6), 1299-1314 (2019-07-30)
MRE11 nuclease forms a trimeric complex (MRN) with RAD50 and NBS1 and plays a central role in preventing genomic instability. When DNA double-strand breaks (DSBs) occur, MRN is quickly recruited to the damage site and initiates DNA end resection; accordingly
Minghao Yang et al.
Cell death & disease, 13(9), 815-815 (2022-09-24)
Osteosarcoma (OS), the most common primary malignancy of the bone, has a poor prognosis due to its high mortality rate and high potential for metastasis. Thus, it is urgently necessary to explore functional molecular targets of therapeutic strategies for osteosarcoma.
Li-Ming Liu et al.
Life (Basel, Switzerland), 11(8) (2021-08-28)
The protein p53 is one of the most important tumor suppressors, responding to a variety of stress signals. Mutations in p53 occur in about half of human cancer cases, and dysregulation of the p53 function by epigenetic modifiers and modifications
Gang Ren et al.
Scientific reports, 9(1), 6688-6688 (2019-05-02)
One third of newly diagnosed breast cancers in the US are early-stage lesions. The etiological understanding and treatment of these lesions have become major clinical challenges. Because breast cancer risk factors are often linked to aberrant nitric oxide (NO) production

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service