Skip to Content
Merck

Skip To

1.10255

Bis(trimethylsilyl)trifluoroacetamide

for gas chromatography

Synonym(s):

Bis(trimethylsilyl)trifluoroacetamide, 2,2,2-Trifluoro-N,O-bis(trimethylsilyl)acetamide

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C8H18F3NOSi2
CAS Number:
Molecular Weight:
257.40
UNSPSC Code:
12352103
EC Index Number:
247-103-9
NACRES:
NA.05
MDL number:
Assay:
≥98.0% (GC)
Form:
liquid


Quality Segment

assay

≥98.0% (GC)

form

liquid

technique(s)

gas chromatography (GC): suitable

bp

145 °C/1013 hPa

transition temp

flash point 34 °C

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](N=C(O[Si](C)(C)C)C(F)(F)F)(C)(C)C

InChI

1S/C8H18F3NOSi2/c1-14(2,3)12-7(8(9,10)11)13-15(4,5)6/h1-6H3

InChI key

XCOBLONWWXQEBS-UHFFFAOYSA-N

Application


  • Determination of Delta(9)-tetrahydrocannabinol, 11-nor-carboxy-Delta(9)-tetrahydrocannabinol and cannabidiol in human plasma and urine after a commercial cannabidiol oil product intake.: This study uses Bis(trimethylsilyl)trifluoroacetamide (BSTFA) for derivatization in the analysis of cannabinoids in biological samples. The method enhances the detection sensitivity and accuracy, which is critical in forensic toxicology and clinical research (Papoutsis et al., 2024).

  • Simultaneous determination of 19 bromophenols by gas chromatography-mass spectrometry (GC-MS) after derivatization.: BSTFA is utilized for the derivatization of bromophenols in environmental samples, enhancing the separation and detection of these compounds in GC-MS analysis. This method is crucial for monitoring environmental contaminants (Liu et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
15222T638115238
assay

≥98.0% (GC)

assay

≥99.0% (GC)

assay

-

assay

≥98.5% (excluding chlorotrimethylsilane, GC), 99% (excluding TMCS)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

density

0.97 g/cm3 at 20 °C

density

0.969 g/mL at 25 °C (lit.)

density

0.97 g/mL (lit.)

density

0.97 g/mL (lit.)

form

liquid

form

liquid

form

-

form

liquid

storage temp.

2-30°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-

bp

145 °C/1013 hPa

bp

45-50 °C/14 mmHg (lit.)

bp

45-50 °C/14 mmHg (lit.)

bp

45-50 °C/14 mmHg (lit.)


pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

93.2 °F

flash_point_c

34 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters