Skip to Content
Merck
All Photos(2)

Key Documents

T6381

Supelco

N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane

N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane
1 of 1 reviewers received a sample product or took part in a promotion

with 1% trimethylchlorosilane, LiChropur

Synonym(s):

BSTFA + TMCS

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
€59.10
5 G
€67.10
25 G
€163.00
10 AMP
€279.00
100 G
€1,000.00

€59.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 AMP
€59.10
5 G
€67.10
25 G
€163.00
10 AMP
€279.00
100 G
€1,000.00

About This Item

Linear Formula:
CF3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
257.40
Beilstein:
2050024
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.22

€59.10


Check Cart for Availability

Request a Bulk Order

grade

derivatization grade ((GC derivatization))

Quality Level

quality

LiChropur
with 1% trimethylchlorosilane

contains

1% trimethylchlorosilane as additive

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

color

hazy clear to colorless

refractive index

n20/D 1.384 (lit.)

bp

45-50 °C/14 mmHg (lit.)

solubility

hexane: miscible 100 mg/mL

density

0.97 g/mL (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)O\C(=N\[Si](C)(C)C)C(F)(F)F

InChI

1S/C8H18F3NOSi2/c1-14(2,3)12-7(8(9,10)11)13-15(4,5)6/h1-6H3/b12-7+

InChI key

XCOBLONWWXQEBS-KPKJPENVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane is a silylating reagent generally used in estrogenic steroid hormone identification and quantification.[1]

Application

BSTFA+TMCS has been used as derivatizing reagent for GC-MS determination of natural and synthetic estrogenic steroids.[1]

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)
Sealed ampules contain 1 ml. Gram sizes are in screw-cap bottles.

Other Notes

The position of the silyl groups has not been unequivocally established. It has been referred to as both N,N and N,O.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.1 °F - closed cup

Flash Point(C)

6.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pitfalls and solution for simultaneous determination of estrone and 17a-ethinylestradiol by gas chromatography?mass spectrometry after derivatization with N, O-bis (trimethylsilyl) trifluoroacetamide.
Zhang, Kai, and Yuegang Zuo.
Analytica Chimica Acta, 554.1, 190-196 (2005)
Shabtai Cohen et al.
Plants (Basel, Switzerland), 9(9) (2020-08-23)
Emerging needs for diversifying human diet and to explore novel therapeutic procedures have led to increasing attempts to retrieve traditional nourishments and recruit beneficial wild plant species. Species of the genus Erodium (Geraniaceae) harbor medicinal indications and substances known from
Hidenori Matsukami et al.
The Science of the total environment, 493, 672-681 (2014-07-06)
Chlorinated and non-halogenated organophosphorus flame retardants (OPFRs) including tris(2-chloroisopropyl) phosphate (TCIPP), diethylene glycol bis(di(2-chloroisopropyl) phosphate) (DEG-BDCIPP), triphenyl phosphate (TPHP), and bisphenol A bis(diphenyl phosphate) (BPA-BDPP) have been used increasingly as alternatives to polybrominated diphenyl ethers and other brominated flame retardants.
Ahmed I Rushdi et al.
SpringerPlus, 3, 253-253 (2014-06-14)
Propolis is a sticky material mixed by honeybees to utilize it in protecting their hives from infection by bacteria and fungi. The therapeutic properties of propolis are due to its chemical composition with bio-active compounds; therefore, researchers are interested in
Nyi Nyi Naing et al.
Journal of chromatography. A, 1423, 1-8 (2015-11-05)
Electro membrane extraction-solid-liquid phase microextraction (EME-SLPME) was developed for the first time to determine phenolic contaminants in water. The extraction system consisted of a solid/liquid interface that permitted a three-phase microextraction approach involving an aqueous sample (donor phase): an organic

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service