Skip to Content
Merck
All Photos(1)

Documents

1.09480

Supelco

ICP multi-element standard solution XIII

(15 elements in diluted nitric acid) Certipur®

Synonym(s):

Nota 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

reference material

Quality Level

product line

Certipur®

technique(s)

ICP: suitable

pH

1 (20 °C in H2O)

density

1.032 g/cm3

application(s)

industrial qc
pharmaceutical

format

multi-component solution

storage temp.

15-25°C

Analysis Note

Al (Aluminium): 500 mg/l
As (arsenic): 100 mg/l
Be (beryllium): 100 mg/l
Cd (cadmium): 25.0 mg/l
Co (cobalt): 100 mg/l
Cr (chromium): 100 mg/l
Cu (Copper): 100 mg/l
Fe (iron): 100 mg/l
Hg (Mercury): 5.0 mg/l
Mn (Manganese): 100 mg/l
Ni (Nickel): 100 mg/l
Pb (lead): 100 mg/l
Se (Selenium): 25.0 mg/l
V (Vanadium): 250 mg/l
Zn (Zinc): 100 mg/l

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

EU REACH Annex XIV (Authorisation List)

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service