Skip to Content
Merck

Skip To

1.15474

ICP multi-element standard solution I

(19 elements in dilute nitric acid) Certipur®

Synonym(s):

Nota 1 A

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107


grade

reference material

Quality Segment

product line

Certipur®

technique(s)

ICP: suitable

pH

0.1 (20 °C in H2O)

density

1.02 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

multi-component solution

storage temp.

15-25°C

Application

ICP multi-element standard solution I can be used as a calibration standard for quantification of 10 major heavy metals , in water-soluble fraction and Polycyclic aromatic hydrocarbon fraction of oil waste samples by inductively coupled plasma-emission spectrometry (ICP-ES).

Analysis Note

Ag (Silver): 50 mg/l
Al (Aluminium): 100 mg/l
B (boron): 15 mg/l
Ba (barium): 5 mg/l
Be (beryllium): 1 mg/l
Bi (bismuth): 200 mg/l
Cd (cadmium): 20 mg/l
Co (cobalt): 20 mg/l
Cr (chromium): 25 mg/l
Cu (Copper): 20 mg/l
Fe (iron): 15 mg/l
Ga (Gallium): 150 mg/l
In (Indium): 200 mg/l
Mn (Manganese): 5 mg/l
Ni (Nickel): 50 mg/l
Pb (lead): 200 mg/l
Sr (Strontium): 1 mg/l
Tl (Thallium): 400 mg/l
Zn (Zinc): 20 mg/l

Other Notes

Blocked in the US and Canada due to regulatory reasons
For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

supp_hazards

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

10325-94-7 + 10099-74-8 + 10141-05-6

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
115474010004022536160481

Questions

Reviews

No rating value

Active Filters