Skip to Content
Merck
All Photos(1)

Key Documents

1.09493

Supelco

ICP multi-element standard solution X

for surface water testing (23 elements in dilute nitric acid) Certipur®

Synonym(s):

Multi-Element Calibration Solution for ICP/AAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

reference material

Quality Level

product line

Certipur®

technique(s)

ICP: suitable

pH

1 (20 °C in H2O)

bp

100 °C/1013 hPa

density

1.007 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

multi-component solution

storage temp.

15-25°C

Related Categories

Application


  • Trace element analysis in bovine semen: This research utilizes inductively coupled plasma mass spectrometry to determine trace elements in bovine semen samples, showcasing the versatility of ICP multi-element standard solutions in diverse biotechnological applications and quality control processes (Aguiar et al., 2012).

  • Development of slurry introduction method for antibiotics: The development of a new slurry introduction method for multi-element analysis of antibiotics highlights the application of ICP multi-element standard solutions in enhancing pharmaceutical research methodologies, particularly in the measurement of elemental composition in complex matrices (Zachariadis and Michos, 2007).



Analysis Note

Ca (calcium): 35000 µg/l
Mg (Magnesium): 15000 µg/l
Na (sodium): 8000 µg/l
K (potassium): 3000 µg/l
B (boron): 100 µg/l
Fe (iron): 100 µg/l
Mo (Molydenium): 100 µg/l
Sr (Strontium): 100 µg/l
As (arsenic): 50 µg/l
Ba (barium): 50 µg/l
Ni (Nickel): 50 µg/l
V (Vanadium): 50 µg/l
Zn (Zinc): 50 µg/l
Mn (Manganese): 30 µg/l
Co (cobalt): 25 µg/l
Pb (lead): 25 µg/l
Be (beryllium): 20 µg/l
Cd (cadmium): 20 µg/l
Cr (chromium): 20 µg/l
Cu (Copper): 20 µg/l
Bi (bismuth): 10 µg/l
Se (Selenium): 10 µg/l
Tl (Thallium): 10 µg/l

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service