Skip to Content
Merck
All Photos(1)

Key Documents

1.10580

Supelco

ICP multi-element standard solution VI

for ICP-MS (30 elements in dilute nitric acid) Certipur®

Synonym(s):

Nota 1 A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

reference material

Quality Level

product line

Certipur®

pH

0.5 (20 °C in H2O)

density

1.03 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

multi-component solution

storage temp.

15-25°C

General description

for ICP-MS (30 elements in dilute nitric acid) Certipur®

Application

ICP multi-element standard solution VI can be used as a calibration standard for the determination of trace elements in hydrogenated fats using inductively coupled plasma-mass spectrometry (ICP MS).[1]

Analysis Note

In 6% HNO3:
  • 10 mg/L: Ag, Al, Ba, Bi, Cd, Co, Cr, Cu, Ga, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Rb, Sr, Te, Tl, U, V
  • 100 mg/L: As, B, Be, Ca, Fe, Zn
  • 1000 mg/L: Ca

Other Notes

Blocked in the US and Canada due to regulatory reasons

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service