Skip to Content
Merck

Skip To

1.06946

LM

antioxidant for fats and oils, EMPROVE® ESSENTIAL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 kg
Check Cart for Availability
€ 411,00

About This Item

UNSPSC Code:
12352401

€ 411,00


Check Cart for Availability

Request a Bulk OrderRequest a Sample
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

product line

EMPROVE® ESSENTIAL

form

paste

mp

55 °C

density

1.04 g/cm3 at 20 °C

application(s)

pharmaceutical
semi-solid formulation

storage temp.

2-8°C

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

LM Emprove® Essential acts as an antioxidant and stabilizer in formulations. LM Emprove® Essential is suitable for pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.083301.071304.80864
LM antioxidant for fats and oils, EMPROVE® ESSENTIAL

SAFC

1.06946

LM

form

paste

form

liquid

form

powder

form

liquid

application(s)

pharmaceutical
semi-solid formulation

application(s)

pharmaceutical
semi-solid formulation

application(s)

pharmaceutical
solid formulation

application(s)

parenterals
pharmaceutical

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® EXPERT

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-30°C

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

500

mp

55 °C

mp

-

mp

-

mp

-


Still not finding the right product?

Explore all of our products under LM


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

hcodes



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
106946100004022536085135

Questions

Reviews

No rating value

Active Filters